missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(4-Acetyl-1-piperazinyl)benzeneboronic acid pinacol ester, 95%, Thermo Scientific Chemicals
123.00 € - 390.00 €
Chemical Identifiers
CAS | 1218791-38-8 |
---|---|
Molecular Formula | C18H27BN2O3 |
Molecular Weight (g/mol) | 330.235 |
MDL Number | MFCD13195756 |
InChI Key | VRZVSHHLWMAHAZ-UHFFFAOYSA-N |
Synonym | 4-4-acetyl-1-piperazinyl phenylboronic acid pinacol ester, 1-4-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl piperazin-1-yl ethanone, 4-4-acetylpiperazino phenylboronic acid pinacol ester, 1-4-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl-1-piperazinyl ethanone, 4-4-acetyl-1-piperazinyl phenylboronicacidpinacolester, 4-4-acetylpiperazino phenylboronic acid, pinacol ester, 4-4-acetyl-1-piperazinyl benzeneboronic acid pinacol ester, 1-4-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl piperazin-1-yl ethan-1-one |
PubChem CID | 53217141 |
IUPAC Name | 1-[4-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]piperazin-1-yl]ethanone |
SMILES | B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)N3CCN(CC3)C(=O)C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15445256
|
Thermo Scientific Alfa Aesar
H59771.MD |
250 mg | N/A | ||||||
15435256
|
Thermo Scientific Alfa Aesar
H59771.03 |
1 g |
390.00 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
NotesStore away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Chemical Identifiers
1218791-38-8 | |
330.235 | |
VRZVSHHLWMAHAZ-UHFFFAOYSA-N | |
53217141 | |
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)N3CCN(CC3)C(=O)C |
C18H27BN2O3 | |
MFCD13195756 | |
4-4-acetyl-1-piperazinyl phenylboronic acid pinacol ester, 1-4-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl piperazin-1-yl ethanone, 4-4-acetylpiperazino phenylboronic acid pinacol ester, 1-4-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl-1-piperazinyl ethanone, 4-4-acetyl-1-piperazinyl phenylboronicacidpinacolester, 4-4-acetylpiperazino phenylboronic acid, pinacol ester, 4-4-acetyl-1-piperazinyl benzeneboronic acid pinacol ester, 1-4-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl piperazin-1-yl ethan-1-one | |
1-[4-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]piperazin-1-yl]ethanone |
Specifications
1218791-38-8 | |
MFCD13195756 | |
VRZVSHHLWMAHAZ-UHFFFAOYSA-N | |
1-[4-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]piperazin-1-yl]ethanone | |
53217141 | |
95% |
C18H27BN2O3 | |
4-4-acetyl-1-piperazinyl phenylboronic acid pinacol ester, 1-4-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl piperazin-1-yl ethanone, 4-4-acetylpiperazino phenylboronic acid pinacol ester, 1-4-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl-1-piperazinyl ethanone, 4-4-acetyl-1-piperazinyl phenylboronicacidpinacolester, 4-4-acetylpiperazino phenylboronic acid, pinacol ester, 4-4-acetyl-1-piperazinyl benzeneboronic acid pinacol ester, 1-4-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl piperazin-1-yl ethan-1-one | |
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)N3CCN(CC3)C(=O)C | |
330.235 | |
330.23 | |
4-(4-Acetyl-1-piperazinyl)benzeneboronic acid pinacol ester |
Safety and Handling
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only