missing translation for 'onlineSavingsMsg'
Learn More

4,4'-Bis(dimethylamino)benzil, 98%, Thermo Scientific Chemicals

45.70 € - 467.00 €

Chemical Identifiers

CAS 17078-27-2
Molecular Formula C18H20N2O2
Molecular Weight (g/mol) 296.36
InChI Key AVFUVYIDYFXFSX-UHFFFAOYSA-N
Synonym 4,4'-bis dimethylamino benzil, 1,2-bis 4-dimethylamino phenyl ethane-1,2-dione, ethandione, bis 4-dimethylamino phenyl, bis 4-dimethylamino phenyl ethane-1,2-dione, acmc-209e1j, benzil-based compound, 35, bis n,n-dimethylamino benzil, 4,4/'-bis dimethylamino benzil, bis 4-dimethylamino phenyl diketone
PubChem CID 222570
IUPAC Name 1,2-bis[4-(dimethylamino)phenyl]ethane-1,2-dione
SMILES CN(C)C1=CC=C(C=C1)C(=O)C(=O)C2=CC=C(C=C2)N(C)C
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10289880
View Documents
Thermo Scientific Acros
402390010
1 g Glass bottle
45.70 €
1g
Please to purchase this item. Need a web account? Register with us today!
10045780
View Documents
Thermo Scientific Acros
402390050
5 g Glass bottle
142.71 €
5g
Please to purchase this item. Need a web account? Register with us today!
11975321
View Documents
Thermo Scientific Acros
402390250
25 g Glass bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

17078-27-2
296.36
4,4'-bis dimethylamino benzil, 1,2-bis 4-dimethylamino phenyl ethane-1,2-dione, ethandione, bis 4-dimethylamino phenyl, bis 4-dimethylamino phenyl ethane-1,2-dione, acmc-209e1j, benzil-based compound, 35, bis n,n-dimethylamino benzil, 4,4/'-bis dimethylamino benzil, bis 4-dimethylamino phenyl diketone
1,2-bis[4-(dimethylamino)phenyl]ethane-1,2-dione
C18H20N2O2
AVFUVYIDYFXFSX-UHFFFAOYSA-N
222570
CN(C)C1=CC=C(C=C1)C(=O)C(=O)C2=CC=C(C=C2)N(C)C

Specifications

17078-27-2
Green to Green-Yellow
97.5% min. (HPLC)
4,4'-bis dimethylamino benzil, 1,2-bis 4-dimethylamino phenyl ethane-1,2-dione, ethandione, bis 4-dimethylamino phenyl, bis 4-dimethylamino phenyl ethane-1,2-dione, acmc-209e1j, benzil-based compound, 35, bis n,n-dimethylamino benzil, 4,4/'-bis dimethylamino benzil, bis 4-dimethylamino phenyl diketone
CN(C)C1=CC=C(C=C1)C(=O)C(=O)C2=CC=C(C=C2)N(C)C
296.36
296.36
Crystalline Powder
200.0°C to 204.0°C
Authentic
C18H20N2O2
AVFUVYIDYFXFSX-UHFFFAOYSA-N
1,2-bis[4-(dimethylamino)phenyl]ethane-1,2-dione
222570
98%
4, 4'-Bis(dimethylamino)benzil
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 4,4'-Bis(dimethylamino)benzil

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only