missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,6-Dimethyl-1,3,5-triazin-2-amine hydrate, ≥95%, Thermo Scientific™
74.10 € - 214.00 €
Chemical Identifiers
CAS | 175278-59-8 |
---|---|
Molecular Formula | C5H10N4O |
Molecular Weight (g/mol) | 142.16 |
MDL Number | MFCD00663061 |
InChI Key | CZKUOCRCKNMNTI-UHFFFAOYSA-N |
Synonym | 4,6-dimethyl-1,3,5-triazin-2-amine hydrate, dimethyl-1,3,5-triazin-2-amine hydrate, 1,3,5-triazin-2-amine,4,6-dimethyl-, hydrate 1:1, 2-amino-4,6-dimethyl-1,3,5-triazine hydrate, 4,6-dimethyl-1,3,5-triazine-2-ylamine, oxamethane, 1,3,5-triazin-2-amine,4,6-dimethyl-,hydrate 1:1 |
PubChem CID | 2782054 |
IUPAC Name | 4,6-dimethyl-1,3,5-triazin-2-amine;hydrate |
SMILES | O.CC1=NC(N)=NC(C)=N1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10334392
|
Thermo Scientific Maybridge
RF02292DA |
1 g | Amber Glass Bottle |
74.10 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10772873
|
Thermo Scientific Maybridge
RF02292EA |
10 g | Amber Glass Bottle |
214.00 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
175278-59-8 | |
142.16 | |
CZKUOCRCKNMNTI-UHFFFAOYSA-N | |
2782054 | |
O.CC1=NC(N)=NC(C)=N1 |
C5H10N4O | |
MFCD00663061 | |
4,6-dimethyl-1,3,5-triazin-2-amine hydrate, dimethyl-1,3,5-triazin-2-amine hydrate, 1,3,5-triazin-2-amine,4,6-dimethyl-, hydrate 1:1, 2-amino-4,6-dimethyl-1,3,5-triazine hydrate, 4,6-dimethyl-1,3,5-triazine-2-ylamine, oxamethane, 1,3,5-triazin-2-amine,4,6-dimethyl-,hydrate 1:1 | |
4,6-dimethyl-1,3,5-triazin-2-amine;hydrate |
Specifications
175278-59-8 | |
95%+ | |
MFCD00663061 | |
CZKUOCRCKNMNTI-UHFFFAOYSA-N | |
4,6-dimethyl-1,3,5-triazin-2-amine;hydrate | |
2782054 | |
95% |
95.0 | |
C5H10N4O | |
4,6-dimethyl-1,3,5-triazin-2-amine hydrate, dimethyl-1,3,5-triazin-2-amine hydrate, 1,3,5-triazin-2-amine,4,6-dimethyl-, hydrate 1:1, 2-amino-4,6-dimethyl-1,3,5-triazine hydrate, 4,6-dimethyl-1,3,5-triazine-2-ylamine, oxamethane, 1,3,5-triazin-2-amine,4,6-dimethyl-,hydrate 1:1 | |
O.CC1=NC(N)=NC(C)=N1 | |
142.16 | |
142.17 | |
4,6-Dimethyl-1,3,5-triazin-2-amine hydrate |