missing translation for 'onlineSavingsMsg'
Learn More

4-(6-Hydrazino-4-pyrimidinyl)morpholine, 96%, Thermo Scientific™

74.30 € - 188.00 €

Chemical Identifiers

CAS 5767-36-2
Molecular Formula C8H13N5O
Molecular Weight (g/mol) 195.226
MDL Number MFCD11655642
InChI Key XWDNLTCJJYYYCI-UHFFFAOYSA-N
Synonym 4-6-hydrazinylpyrimidin-4-yl morpholine, 4-6-hydrazinopyrimidin-4-yl morpholine, morpholine, 4-6-hydrazinyl-4-pyrimidinyl, 4-6-hydrazino-4-pyrimidinyl morpholine, 6-morpholin-4-ylpyrimidine-4-ylhydrazine, 4-6-hydrazino-pyrimidin-4-yl-morpholine, 6-morpholin-4-yl-pyrimidin-4-yl-hydrazine
PubChem CID 13393622
IUPAC Name (6-morpholin-4-ylpyrimidin-4-yl)hydrazine
SMILES C1COCCN1C2=CC(=NC=N2)NN
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15486427
View Documents
Thermo Scientific Alfa Aesar
H33589.MD
250 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15476427
View Documents
Thermo Scientific Alfa Aesar
H33589.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

5767-36-2
195.226
XWDNLTCJJYYYCI-UHFFFAOYSA-N
13393622
C1COCCN1C2=CC(=NC=N2)NN
C8H13N5O
MFCD11655642
4-6-hydrazinylpyrimidin-4-yl morpholine, 4-6-hydrazinopyrimidin-4-yl morpholine, morpholine, 4-6-hydrazinyl-4-pyrimidinyl, 4-6-hydrazino-4-pyrimidinyl morpholine, 6-morpholin-4-ylpyrimidine-4-ylhydrazine, 4-6-hydrazino-pyrimidin-4-yl-morpholine, 6-morpholin-4-yl-pyrimidin-4-yl-hydrazine
(6-morpholin-4-ylpyrimidin-4-yl)hydrazine

Specifications

5767-36-2
C8H13N5O
4-6-hydrazinylpyrimidin-4-yl morpholine, 4-6-hydrazinopyrimidin-4-yl morpholine, morpholine, 4-6-hydrazinyl-4-pyrimidinyl, 4-6-hydrazino-4-pyrimidinyl morpholine, 6-morpholin-4-ylpyrimidine-4-ylhydrazine, 4-6-hydrazino-pyrimidin-4-yl-morpholine, 6-morpholin-4-yl-pyrimidin-4-yl-hydrazine
C1COCCN1C2=CC(=NC=N2)NN
195.226
195.22
4-(6-Hydrazino-4-pyrimidinyl)morpholine
162°C to 163°C
MFCD11655642
XWDNLTCJJYYYCI-UHFFFAOYSA-N
(6-morpholin-4-ylpyrimidin-4-yl)hydrazine
13393622
96%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c

H302+H312+H332

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.