missing translation for 'onlineSavingsMsg'
Learn More
4,7-Diphenyl-1,10-phenanthroline, 99%, Thermo Scientific Chemicals Product Code.: 10696932

4,7-Diphenyl-1,10-phenanthroline, 99%, Thermo Scientific Chemicals

Product Code. 10696932
1 g, Glass bottle
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10696932

Brand: Thermo Scientific Acros 158320010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1662-01-7
Molecular Formula C24H16N2
Molecular Weight (g/mol) 332.39
MDL Number MFCD00004976
InChI Key DHDHJYNTEFLIHY-UHFFFAOYSA-N
Synonym bathophenanthroline, 1,10-phenanthroline, 4,7-diphenyl, bphen, 1,10-bathophenanthroline, bathophenanthrolin, bathophenanthrolin german, unii-4a2b091f0g, 4,7-diphenyl-1,10-diazaphenanthrene, gnf-pf-4554, 4,7-diphenylpyridino 3,2-h quinolineShow MoreShow Less
PubChem CID 72812
ChEBI CHEBI:77995
IUPAC Name 4,7-diphenyl-1,10-phenanthroline
SMILES C1=CC=C(C=C1)C2=C3C=CC4=C(C=CN=C4C3=NC=C2)C5=CC=CC=C5

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 218.0°C to 221.0°C
Color Pink to White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: insoluble.
Formula Weight 332.39
Percent Purity 99%
Physical Form Crystalline Powder, Crystals or Crystalline Mass
Chemical Name or Material 4, 7-Diphenyl-1, 10-phenanthroline
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0467420 Certificate TypeCertificate of Analysis Date30/01/2025 Catalog Number158320010,158320250,158320050
Lot NumberA0465205 Certificate TypeCertificate of Analysis Date01/11/2024 Catalog Number158320010,158320250,158320050
Lot NumberA0420303 Certificate TypeCertificate of Analysis Date05/01/2023 Catalog Number158320010,158320250,158320050
Lot NumberA0409395 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number158320010,158320250,158320050
Lot NumberA0412605 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number158320010,158320250,158320050

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4,7-Diphenyl-1,10-phenanthroline, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.