missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Acetoxy-3-methoxycinnamic acid, 98+%, Thermo Scientific Chemicals
30.90 € - 423.00 €
Chemical Identifiers
CAS | 2596-47-6 |
---|---|
Molecular Formula | C12H12O5 |
Molecular Weight (g/mol) | 236.223 |
MDL Number | MFCD00014378 |
InChI Key | IHKNVZISLLDMOR-GQCTYLIASA-N |
Synonym | acetylferulic acid, 4-acetoxy-3-methoxycinnamic acid, 3-methoxy-4-acetoxycinnamic acid, 3-4-acetoxy-3-methoxyphenyl acrylic acid, cinnamic acid, 4-acetoxy-3-methoxy, acetylated ferulic acid, cinnamic acid, 4-hydroxy-3-methoxy-, acetate, 2-propenoic acid, 3-4-acetyloxy-3-methoxyphenyl, 2e-3-4-acetyloxy-3-methoxyphenyl prop-2-enoic acid |
PubChem CID | 5354677 |
ChEBI | CHEBI:86582 |
IUPAC Name | (E)-3-(4-acetyloxy-3-methoxyphenyl)prop-2-enoic acid |
SMILES | CC(=O)OC1=C(C=C(C=C1)C=CC(=O)O)OC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11444163
|
Thermo Scientific Alfa Aesar
A10361.03 |
1 g |
30.90 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11454163
|
Thermo Scientific Alfa Aesar
A10361.06 |
5 g |
107.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11464163
|
Thermo Scientific Alfa Aesar
A10361.14 |
25 g |
423.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
2596-47-6 | |
236.223 | |
IHKNVZISLLDMOR-GQCTYLIASA-N | |
5354677 | |
(E)-3-(4-acetyloxy-3-methoxyphenyl)prop-2-enoic acid |
C12H12O5 | |
MFCD00014378 | |
acetylferulic acid, 4-acetoxy-3-methoxycinnamic acid, 3-methoxy-4-acetoxycinnamic acid, 3-4-acetoxy-3-methoxyphenyl acrylic acid, cinnamic acid, 4-acetoxy-3-methoxy, acetylated ferulic acid, cinnamic acid, 4-hydroxy-3-methoxy-, acetate, 2-propenoic acid, 3-4-acetyloxy-3-methoxyphenyl, 2e-3-4-acetyloxy-3-methoxyphenyl prop-2-enoic acid | |
CHEBI:86582 | |
CC(=O)OC1=C(C=C(C=C1)C=CC(=O)O)OC |
Specifications
2596-47-6 | |
Odorless | |
MFCD00014378 | |
acetylferulic acid, 4-acetoxy-3-methoxycinnamic acid, 3-methoxy-4-acetoxycinnamic acid, 3-4-acetoxy-3-methoxyphenyl acrylic acid, cinnamic acid, 4-acetoxy-3-methoxy, acetylated ferulic acid, cinnamic acid, 4-hydroxy-3-methoxy-, acetate, 2-propenoic acid, 3-4-acetyloxy-3-methoxyphenyl, 2e-3-4-acetyloxy-3-methoxyphenyl prop-2-enoic acid | |
CC(=O)OC1=C(C=C(C=C1)C=CC(=O)O)OC | |
236.223 | |
CHEBI:86582 | |
≥98% |
193°C to 196°C | |
C12H12O5 | |
2218026 | |
IHKNVZISLLDMOR-GQCTYLIASA-N | |
(E)-3-(4-acetyloxy-3-methoxyphenyl)prop-2-enoic acid | |
5354677 | |
236.22 | |
4-Acetoxy-3-methoxycinnamic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only