missing translation for 'onlineSavingsMsg'
Learn More
4-Aminoazobenzene-4'-sulfonic acid sodium salt, tech. 90%, Thermo Scientific Chemicals Product Code.: 11378525

4-Aminoazobenzene-4'-sulfonic acid sodium salt, tech. 90%, Thermo Scientific Chemicals

Product Code. 11378525
100 g
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy

Product Code. 11378525

Brand: Thermo Scientific Alfa Aesar L11965.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2491-71-6
Molecular Formula C12H10N3NaO3S
Molecular Weight (g/mol) 299.28
MDL Number MFCD00035564
InChI Key FIXVWFINKCQNFG-UHFFFAOYSA-M
Synonym yellow rfs, 11648 yellow, acid yellow fwa, new yellow gmf, 4-aminoazobenzene-4'-sulfonic acid sodium salt, hexacol yellow rfs, c.i. food yellow 6, monosodium salt, sodium 4'-aminoazobenzene-4-sulphonate, yellow r.i.s., unii-z452e7h2u8Show MoreShow Less
PubChem CID 23691997
IUPAC Name sodium;4-[(4-aminophenyl)diazenyl]benzenesulfonate
SMILES C1=CC(=CC=C1N)N=NC2=CC=C(C=C2)S(=O)(=O)[O-].[Na+]

Specifications

Quantity 100 g
Beilstein 8098548
Sensitivity Very Hygroscopic
Formula Weight 299.29
Percent Purity 90%
Grade Technical
Chemical Name or Material 4-Aminoazobenzene-4'-sulfonic acid sodium salt

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10253494 Certificate TypeCertificate of Analysis Date31/12/2024 Catalog NumberL11965.22,L11965.14
Lot Number10252605 Certificate TypeCertificate of Analysis Date10/10/2024 Catalog NumberL11965.22,L11965.14
Lot Number10250283 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberL11965.22,L11965.14
Lot Number10251477 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberL11965.22,L11965.14
Lot Number10251772 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberL11965.22,L11965.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Aminoazobenzene-4'-sulfonic acid sodium salt, tech. 90%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.