Learn More
4-Benzyloxy-3-chlorobenzeneboronic acid, 98%, Thermo Scientific™
20.50 € - 64.70 €
Chemical Identifiers
CAS | 845551-44-2 |
---|---|
Molecular Formula | C13H12BClO3 |
Molecular Weight (g/mol) | 262.496 |
MDL Number | MFCD04115642 |
InChI Key | AYSMMNDQVZAXQY-UHFFFAOYSA-N |
Synonym | 4-benzyloxy-3-chlorophenylboronic acid, 4-benzyloxy-3-chlorophenyl boronic acid, 3-chloro-4-benzyloxyphenyl boronic acid, 4-benzyloxy-3-chlorobenzeneboronic acid, boronic acid, 3-chloro-4-phenylmethoxy phenyl, b-3-chloro-4-phenylmethoxy phenyl-boronic acid, akos brn-0594, acmc-209pvk, 4-benzyloxy-3-chlorophenylboro |
PubChem CID | 3763657 |
IUPAC Name | (3-chloro-4-phenylmethoxyphenyl)boronic acid |
SMILES | B(C1=CC(=C(C=C1)OCC2=CC=CC=C2)Cl)(O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15429807
|
Thermo Scientific Alfa Aesar
H53033.03 |
1 g | N/A | ||||||
15439807
|
Thermo Scientific Alfa Aesar
H53033.06 |
5 g |
64.70 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
845551-44-2 | |
262.496 | |
AYSMMNDQVZAXQY-UHFFFAOYSA-N | |
3763657 | |
B(C1=CC(=C(C=C1)OCC2=CC=CC=C2)Cl)(O)O |
C13H12BClO3 | |
MFCD04115642 | |
4-benzyloxy-3-chlorophenylboronic acid, 4-benzyloxy-3-chlorophenyl boronic acid, 3-chloro-4-benzyloxyphenyl boronic acid, 4-benzyloxy-3-chlorobenzeneboronic acid, boronic acid, 3-chloro-4-phenylmethoxy phenyl, b-3-chloro-4-phenylmethoxy phenyl-boronic acid, akos brn-0594, acmc-209pvk, 4-benzyloxy-3-chlorophenylboro | |
(3-chloro-4-phenylmethoxyphenyl)boronic acid |
Specifications
845551-44-2 | |
C13H12BClO3 | |
4-benzyloxy-3-chlorophenylboronic acid, 4-benzyloxy-3-chlorophenyl boronic acid, 3-chloro-4-benzyloxyphenyl boronic acid, 4-benzyloxy-3-chlorobenzeneboronic acid, boronic acid, 3-chloro-4-phenylmethoxy phenyl, b-3-chloro-4-phenylmethoxy phenyl-boronic acid, akos brn-0594, acmc-209pvk, 4-benzyloxy-3-chlorophenylboro | |
B(C1=CC(=C(C=C1)OCC2=CC=CC=C2)Cl)(O)O | |
262.496 | |
262.5 | |
4-Benzyloxy-3-chlorobenzeneboronic acid |
154°C to 162°C | |
MFCD04115642 | |
AYSMMNDQVZAXQY-UHFFFAOYSA-N | |
(3-chloro-4-phenylmethoxyphenyl)boronic acid | |
3763657 | |
98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
Your input is important to us. Please complete this form to provide feedback related to the content on this product.