Learn More
4-Benzyloxy-3-fluorobenzeneboronic acid, 98%, Thermo Scientific™
35.70 € - 83.20 €
Chemical Identifiers
CAS | 133057-83-7 |
---|---|
Molecular Formula | C13H12BFO3 |
Molecular Weight (g/mol) | 246.044 |
MDL Number | MFCD00994627 |
InChI Key | DQPDLDQJMUGVGI-UHFFFAOYSA-N |
Synonym | 4-benzyloxy-3-fluorophenylboronic acid, 3-fluoro-4-benzyloxyphenyl boronic acid, 4-benzyloxy-3-fluorobenzeneboronic acid, 4-benzyloxy-3-fluorophenyl boronic acid, 4-benzyloxy-3-fluorophenyl boranediol, boronic acid, 3-fluoro-4-phenylmethoxy phenyl, pubchem5322 |
PubChem CID | 2737788 |
IUPAC Name | (3-fluoro-4-phenylmethoxyphenyl)boronic acid |
SMILES | B(C1=CC(=C(C=C1)OCC2=CC=CC=C2)F)(O)O |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
133057-83-7 | |
246.044 | |
DQPDLDQJMUGVGI-UHFFFAOYSA-N | |
2737788 | |
B(C1=CC(=C(C=C1)OCC2=CC=CC=C2)F)(O)O |
C13H12BFO3 | |
MFCD00994627 | |
4-benzyloxy-3-fluorophenylboronic acid, 3-fluoro-4-benzyloxyphenyl boronic acid, 4-benzyloxy-3-fluorobenzeneboronic acid, 4-benzyloxy-3-fluorophenyl boronic acid, 4-benzyloxy-3-fluorophenyl boranediol, boronic acid, 3-fluoro-4-phenylmethoxy phenyl, pubchem5322 | |
(3-fluoro-4-phenylmethoxyphenyl)boronic acid |
Specifications
133057-83-7 | |
C13H12BFO3 | |
4-benzyloxy-3-fluorophenylboronic acid, 3-fluoro-4-benzyloxyphenyl boronic acid, 4-benzyloxy-3-fluorobenzeneboronic acid, 4-benzyloxy-3-fluorophenyl boronic acid, 4-benzyloxy-3-fluorophenyl boranediol, boronic acid, 3-fluoro-4-phenylmethoxy phenyl, pubchem5322 | |
B(C1=CC(=C(C=C1)OCC2=CC=CC=C2)F)(O)O | |
246.044 | |
246.04 | |
4-Benzyloxy-3-fluorobenzeneboronic acid |
129°C to 131°C | |
MFCD00994627 | |
DQPDLDQJMUGVGI-UHFFFAOYSA-N | |
(3-fluoro-4-phenylmethoxyphenyl)boronic acid | |
2737788 | |
98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 000-000-0
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures