missing translation for 'onlineSavingsMsg'
Learn More

4-Benzylpiperazine-1-carboximidamide hydroiodide, 95%, Thermo Scientific™

Chemical Identifiers

CAS 852228-15-0
Molecular Formula C12H19IN4
Molecular Weight (g/mol) 346.22
MDL Number MFCD09025921
InChI Key CXQITJKGJPFOIV-UHFFFAOYSA-N
Synonym 4-benzylpiperazine-1-carboximidamide hydroiodide, 4-benzylpiperazine-1-carboximidamide-hydrogen iodide 1/1
PubChem CID 18525929
IUPAC Name 4-benzylpiperazine-1-carboximidamide;hydroiodide
SMILES I.NC(=N)N1CCN(CC2=CC=CC=C2)CC1
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10387644
View Documents Promotion Details
Thermo Scientific Maybridge
MO07679DA
1 g Amber Glass Bottle N/A
N/A
11316895
View Documents Promotion Details
Thermo Scientific Maybridge
MO07679EA
10 g Amber Glass Bottle N/A
N/A
Description

Description

Specifications

Chemical Identifiers

852228-15-0
346.22
CXQITJKGJPFOIV-UHFFFAOYSA-N
18525929
I.NC(=N)N1CCN(CC2=CC=CC=C2)CC1
C12H19IN4
MFCD09025921
4-benzylpiperazine-1-carboximidamide hydroiodide, 4-benzylpiperazine-1-carboximidamide-hydrogen iodide 1/1
4-benzylpiperazine-1-carboximidamide;hydroiodide

Specifications

852228-15-0
95%
MFCD09025921
CXQITJKGJPFOIV-UHFFFAOYSA-N
4-benzylpiperazine-1-carboximidamide;hydroiodide
18525929
95%
95.0
C12H19IN4
4-benzylpiperazine-1-carboximidamide hydroiodide, 4-benzylpiperazine-1-carboximidamide-hydrogen iodide 1/1
I.NC(=N)N1CCN(CC2=CC=CC=C2)CC1
346.22
346.21
4-Benzylpiperazine-1-carboximidamide hydroiodide
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Harmful in contact with skin.
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents

Product Certifications