Learn More
4-Bromo-2-cyanopyridine, 97%, Thermo Scientific Chemicals
68.10 € - 192.00 €
Chemical Identifiers
CAS | 62150-45-2 |
---|---|
Molecular Formula | C6H3BrN2 |
Molecular Weight (g/mol) | 183.008 |
MDL Number | MFCD04065805 |
InChI Key | CZXDCTUSFIKLIJ-UHFFFAOYSA-N |
PubChem CID | 693283 |
IUPAC Name | 4-bromopyridine-2-carbonitrile |
SMILES | C1=CN=C(C=C1Br)C#N |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15407377
|
Thermo Scientific Alfa Aesar
H34424.MD |
250 mg | N/A | ||||||
15497367
|
Thermo Scientific Alfa Aesar
H34424.03 |
1 g |
192.00 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
62150-45-2 | |
183.008 | |
CZXDCTUSFIKLIJ-UHFFFAOYSA-N | |
4-bromopyridine-2-carbonitrile |
C6H3BrN2 | |
MFCD04065805 | |
693283 | |
C1=CN=C(C=C1Br)C#N |
Specifications
62150-45-2 | |
C6H3BrN2 | |
UN3439 | |
CZXDCTUSFIKLIJ-UHFFFAOYSA-N | |
4-bromopyridine-2-carbonitrile | |
693283 | |
97% |
87°C to 91°C | |
MFCD04065805 | |
4-bromo-2-cyanopyridine, 4-bromopicolinonitrile, 4-bromo-pyridine-2-carbonitrile, 2-pyridinecarbonitrile, 4-bromo, 2-cyano-4-bromopyridine, 4-bromo-2-pyridinecarbonitrile, pubchem23829, 4-bromo-2-cyano-pyridine, 2-pyridinecarbonitrile,4-bromo, abbypharma ap-17-5586 | |
C1=CN=C(C=C1Br)C#N | |
183.008 | |
183.01 | |
4-Bromo-2-cyanopyridine |
Safety and Handling
GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only