missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-bromo-3,5-dimethyl-1-phenyl-1h-pyrazole, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
10 g
Specifications
Specifications
Assay Percent Range | 97% |
Quantity | 250 mg |
Synonym | 4-bromo-3,5-dimethyl-1-phenyl-1h-pyrazole, 1h-pyrazole,4-bromo-3,5-dimethyl-1-phenyl, 1-phenyl-3,5-dimethyl-4-bromo-1h-pyrazole, 1h-pyrazole, 4-bromo-3,5-dimethyl-1-phenyl |
Formula Weight | 251.13 |
Percent Purity | 97% |
Chemical Name or Material | 4-Bromo-3, 5-dimethyl-1-phenyl-1H-pyrazole |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
4-bromo-3,5-dimethyl-1-phenyl-1h-pyrazole, 97%, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction