missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-bromo-3,5-dimethyl-1-phenyl-1h-pyrazole, 97%, Thermo Scientific™
53.10 € - 367.00 €
Chemical Identifiers
CAS | 51294-75-8 |
---|---|
Molecular Formula | C11H11BrN2 |
Molecular Weight (g/mol) | 251.127 |
InChI Key | GNXWITGSOFQXDG-UHFFFAOYSA-N |
PubChem CID | 7039162 |
IUPAC Name | 4-bromo-3,5-dimethyl-1-phenylpyrazole |
SMILES | CC1=C(C(=NN1C2=CC=CC=C2)C)Br |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10035962
|
Thermo Scientific Maybridge
CC48010CB |
250 mg |
53.10 €
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10797475
|
Thermo Scientific Maybridge
CC48010DA |
1 g |
145.00 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10644195
|
Thermo Scientific Maybridge
CC48010EA |
10 g |
367.00 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
51294-75-8 | |
251.127 | |
7039162 | |
CC1=C(C(=NN1C2=CC=CC=C2)C)Br |
C11H11BrN2 | |
GNXWITGSOFQXDG-UHFFFAOYSA-N | |
4-bromo-3,5-dimethyl-1-phenylpyrazole |
Specifications
51294-75-8 | |
C11H11BrN2 | |
GNXWITGSOFQXDG-UHFFFAOYSA-N | |
4-bromo-3,5-dimethyl-1-phenylpyrazole | |
7039162 | |
97% |
97% | |
4-bromo-3,5-dimethyl-1-phenyl-1h-pyrazole, 1h-pyrazole,4-bromo-3,5-dimethyl-1-phenyl, 1-phenyl-3,5-dimethyl-4-bromo-1h-pyrazole, 1h-pyrazole, 4-bromo-3,5-dimethyl-1-phenyl | |
CC1=C(C(=NN1C2=CC=CC=C2)C)Br | |
251.127 | |
251.13 | |
4-Bromo-3, 5-dimethyl-1-phenyl-1H-pyrazole |