missing translation for 'onlineSavingsMsg'
Learn More
4-Bromobenzoic acid, 97%, Thermo Scientific Chemicals Product Code.: 11414174

4-Bromobenzoic acid, 97%, Thermo Scientific Chemicals

Product Code. 11414174
25 g
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy

Product Code. 11414174

Brand: Thermo Scientific Alfa Aesar A12518.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

4-Bromobenzoic acid was used to study the metabolic fate of 2-,3-and 4-bromo benzoic acids in rat hepatocytes incubation using high temperature liquid chromatography. It was used in bromine-specific detection of the metabolites of 2-,3-and 4-bromobenzoic acid in the urine and bile of rats by inductively coupled plasma mass spectrometry. It is also used as an intermediate for the synthesis of agrochemicals, pharmaceutical, chemical intermediates and liquid crystals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromobenzoic acid was used to study the metabolic fate of 2-,3-and 4-bromo benzoic acids in rat hepatocytes incubation using high temperature liquid chromatography. It was used in bromine-specific detection of the metabolites of 2-,3-and 4-bromobenzoic acid in the urine and bile of rats by inductively coupled plasma mass spectrometry. It is also used as an intermediate for the synthesis of agrochemicals, pharmaceutical, chemical intermediates and liquid crystals.

Solubility
Slightly soluble in hot water. Soluble in alcohol and ether.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 586-76-5
Molecular Formula C7H5BrO2
Molecular Weight (g/mol) 201.02
MDL Number MFCD00002529
InChI Key TUXYZHVUPGXXQG-UHFFFAOYSA-N
Synonym p-bromobenzoic acid, benzoic acid, 4-bromo, p-carboxybromobenzene, benzoic acid, p-bromo, p-bromobenzenecarboxylic acid, 4-bromo-benzoic acid, 4-brom-benzoesaeure, 4-carboxybromobenzene, para bromo benzoic acid, 4bromo-benzoic acidShow MoreShow Less
PubChem CID 11464
ChEBI CHEBI:60698
IUPAC Name 4-bromobenzoic acid
SMILES OC(=O)C1=CC=C(Br)C=C1

Specifications

Melting Point 253°C to 258°C
Density 1.894
Odor Odorless
Quantity 25 g
Beilstein 1906923
Merck Index 14,1408
Solubility Information Slightly soluble in hot water. Soluble in alcohol and ether.
Formula Weight 201.02
Percent Purity ≥98%
Chemical Name or Material 4-Bromobenzoic acid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10252255 Certificate TypeCertificate of Analysis Date06/09/2024 Catalog NumberA12518.14,A12518.36,A12518.22
Lot Number10249589 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog NumberA12518.14,A12518.36,A12518.22
Lot Number10248925 Certificate TypeCertificate of Analysis Date18/12/2023 Catalog NumberA12518.14,A12518.36,A12518.22
Lot Number10244863 Certificate TypeCertificate of Analysis Date24/10/2023 Catalog NumberA12518.14,A12518.36,A12518.22
Lot Number10193296 Certificate TypeCertificate of Analysis Date11/01/2023 Catalog NumberA12518.14,A12518.36,A12518.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Bromobenzoic acid, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.