Learn More
4-Bromobenzoic acid, 97%, Thermo Scientific Chemicals
Description
4-Bromobenzoic acid was used to study the metabolic fate of 2-,3-and 4-bromo benzoic acids in rat hepatocytes incubation using high temperature liquid chromatography. It was used in bromine-specific detection of the metabolites of 2-,3-and 4-bromobenzoic acid in the urine and bile of rats by inductively coupled plasma mass spectrometry. It is also used as an intermediate for the synthesis of agrochemicals, pharmaceutical, chemical intermediates and liquid crystals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
4-Bromobenzoic acid was used to study the metabolic fate of 2-,3-and 4-bromo benzoic acids in rat hepatocytes incubation using high temperature liquid chromatography. It was used in bromine-specific detection of the metabolites of 2-,3-and 4-bromobenzoic acid in the urine and bile of rats by inductively coupled plasma mass spectrometry. It is also used as an intermediate for the synthesis of agrochemicals, pharmaceutical, chemical intermediates and liquid crystals.
Solubility
Slightly soluble in hot water. Soluble in alcohol and ether.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Specifications
Specifications
Melting Point | 253°C to 258°C |
Density | 1.894 |
Odor | Odorless |
Quantity | 500 g |
Beilstein | 1906923 |
Merck Index | 14,1408 |
Solubility Information | Slightly soluble in hot water. Soluble in alcohol and ether. |
Formula Weight | 201.02 |
Percent Purity | ≥98% |
Chemical Name or Material | 4-Bromobenzoic acid |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
10252255 | Certificate of Analysis | 06/09/2024 | A12518.14,A12518.36,A12518.22 |
10249589 | Certificate of Analysis | 11/06/2024 | A12518.14,A12518.36,A12518.22 |
10248925 | Certificate of Analysis | 18/12/2023 | A12518.14,A12518.36,A12518.22 |
10244863 | Certificate of Analysis | 24/10/2023 | A12518.14,A12518.36,A12518.22 |
10193296 | Certificate of Analysis | 11/01/2023 | A12518.14,A12518.36,A12518.22 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.