missing translation for 'onlineSavingsMsg'
Learn More
4-Bromodiphenyl ether, 99%, Thermo Scientific Chemicals Product Code.: 11371287

4-Bromodiphenyl ether, 99%, Thermo Scientific Chemicals

Product Code. 11371287
100 g
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy

Product Code. 11371287

Brand: Thermo Scientific Alfa Aesar L01473.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 101-55-3
Molecular Formula C12H9BrO
Molecular Weight (g/mol) 249.107
MDL Number MFCD00000094
InChI Key JDUYPUMQALQRCN-UHFFFAOYSA-N
Synonym 4-bromodiphenyl ether, 4-bromophenyl phenyl ether, 4-bromophenoxybenzene, p-bromodiphenyl ether, p-phenoxybromobenzene, benzene, 1-bromo-4-phenoxy, p-bromophenyl phenyl ether, 4-bromodiphenylether, p-bromophenoxybenzene, ether, p-bromophenyl phenylShow MoreShow Less
PubChem CID 7565
ChEBI CHEBI:77421
IUPAC Name 1-bromo-4-phenoxybenzene
SMILES C1=CC=C(C=C1)OC2=CC=C(C=C2)Br

Specifications

Melting Point 17°C to 18°C
Density 1.42
Boiling Point 304°C to 305°C
Flash Point >110°C (230°F)
Refractive Index 1.608
Quantity 100 g
UN Number UN3082
Beilstein 1308972
Formula Weight 249.11
Percent Purity 99%
Chemical Name or Material 4-Bromodiphenyl ether
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10188591 Certificate TypeCertificate of Analysis Date16/04/2024 Catalog NumberL01473.14,L01473.22
Lot Number10243784 Certificate TypeCertificate of Analysis Date13/10/2022 Catalog NumberL01473.14,L01473.22
Lot Number10147200 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL01473.14,L01473.22
Lot Number5001P06S Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL01473.14,L01473.22
Lot Number81301178 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL01473.14,L01473.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Bromodiphenyl ether, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.