Learn More
(4-Carboxybutyl)triphenylphosphonium bromide, 98%, Thermo Scientific Chemicals
46.23 € - 377.88 €
Chemical Identifiers
CAS | 17814-85-6 |
---|---|
Molecular Formula | C23H24BrO2P |
Molecular Weight (g/mol) | 443.32 |
MDL Number | MFCD00011906 |
InChI Key | MLOSJPZSZWUDSK-UHFFFAOYSA-N |
Synonym | 4-carboxybutyl triphenylphosphonium bromide, phosphonium, 4-carboxybutyl triphenyl-, bromide, 4-carboxybutyl triphenylphosphanium bromide, carboxybutyltriphenylphosphonium bromide, 5-triphenylphosphonio pentanoic acid bromide, 4-carboxybutyl-triphenylphosphonium bromide, triphenyl 4-carboxybutyl phosphonium bromide, 4-carboxybutyl triphenyl phosphonium bromide |
PubChem CID | 161236 |
IUPAC Name | 4-carboxybutyl(triphenyl)phosphanium;bromide |
SMILES | [Br-].OC(=O)CCCC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11429882
|
Thermo Scientific Alfa Aesar
A12023.14 |
25 g |
46.23 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11439882
|
Thermo Scientific Alfa Aesar
A12023.22 |
100 g |
110.55 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11449882
|
Thermo Scientific Alfa Aesar
A12023.36 |
500 g |
377.88 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Phosphonium salt is a key intermediate for manufacture of some types of prostaglandins, which are subsequently used as active ingredients of both human and veterinary drugs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsPhosphonium salt is a key intermediate for manufacture of some types of prostaglandins, which are subsequently used as active ingredients of both human and veterinary drugs.
Solubility
Soluble in ethanol, methanol and soluble water. Insoluble in toluene and hexane.
Notes
Hygroscopic. Store in dry place. Keep away from oxidizing agents.
Chemical Identifiers
17814-85-6 | |
443.32 | |
MLOSJPZSZWUDSK-UHFFFAOYSA-N | |
161236 | |
[Br-].OC(=O)CCCC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 |
C23H24BrO2P | |
MFCD00011906 | |
4-carboxybutyl triphenylphosphonium bromide, phosphonium, 4-carboxybutyl triphenyl-, bromide, 4-carboxybutyl triphenylphosphanium bromide, carboxybutyltriphenylphosphonium bromide, 5-triphenylphosphonio pentanoic acid bromide, 4-carboxybutyl-triphenylphosphonium bromide, triphenyl 4-carboxybutyl phosphonium bromide, 4-carboxybutyl triphenyl phosphonium bromide | |
4-carboxybutyl(triphenyl)phosphanium;bromide |
Specifications
17814-85-6 | |
195°C (383°F) | |
HO2C(CH2)4P(C6H5)3Br | |
3586477 | |
4-carboxybutyl triphenylphosphonium bromide, phosphonium, 4-carboxybutyl triphenyl-, bromide, 4-carboxybutyl triphenylphosphanium bromide, carboxybutyltriphenylphosphonium bromide, 5-triphenylphosphonio pentanoic acid bromide, 4-carboxybutyl-triphenylphosphonium bromide, triphenyl 4-carboxybutyl phosphonium bromide, 4-carboxybutyl triphenyl phosphonium bromide | |
MLOSJPZSZWUDSK-UHFFFAOYSA-N | |
4-carboxybutyl(triphenyl)phosphanium;bromide | |
161236 | |
98% |
205°C to 209°C | |
C23H24BrO2P | |
MFCD00011906 | |
Hygroscopic | |
Soluble in ethanol,methanol and soluble water. Insoluble in toluene and hexane. | |
[Br-].OC(=O)CCCC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 | |
443.32 | |
443.33 | |
(4-Carboxybutyl)triphenylphosphonium bromide |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
missing translation for 'einecsNumber' : 241-782-5
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only