Learn More
4-Chlorobenzaldehyde, 98%, Thermo Scientific Chemicals
27.64 € - 300.00 €
Chemical Identifiers
CAS | 104-88-1 |
---|---|
Molecular Formula | C7H5ClO |
Molecular Weight (g/mol) | 140.57 |
InChI Key | AVPYQKSLYISFPO-UHFFFAOYSA-N |
Synonym | p-chlorobenzaldehyde, benzaldehyde, 4-chloro, benzaldehyde, p-chloro, p-chlorobenzenecarboxaldehyde, 4-chloro-benzaldehyde, 4-chlorobenzenaldehyde, 4-chloro benzaldehyde, ccris 857, para-chlorobenzaldehyde, unii-e67727up9z |
PubChem CID | 7726 |
ChEBI | CHEBI:28105 |
IUPAC Name | 4-chlorobenzaldehyde |
SMILES | C1=CC(=CC=C1C=O)Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10357560
|
Thermo Scientific Acros
150441000 |
100 g | Glass bottle |
27.64 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10397700
|
Thermo Scientific Acros
150442500 |
250 g | Glass bottle |
35.38 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10523701
|
Thermo Scientific Acros
150440010 |
1 kg | Glass bottle |
96.48 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11944801
|
Thermo Scientific Acros
150440050 |
5 kg | Plastic drum | N/A | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
104-88-1 | |
140.57 | |
p-chlorobenzaldehyde, benzaldehyde, 4-chloro, benzaldehyde, p-chloro, p-chlorobenzenecarboxaldehyde, 4-chloro-benzaldehyde, 4-chlorobenzenaldehyde, 4-chloro benzaldehyde, ccris 857, para-chlorobenzaldehyde, unii-e67727up9z | |
CHEBI:28105 | |
C1=CC(=CC=C1C=O)Cl |
C7H5ClO | |
AVPYQKSLYISFPO-UHFFFAOYSA-N | |
7726 | |
4-chlorobenzaldehyde |
Specifications
104-88-1 | |
White to Yellow | |
214.0°C | |
Authentic | |
C7H5ClO | |
07,235 | |
p-chlorobenzaldehyde, benzaldehyde, 4-chloro, benzaldehyde, p-chloro, p-chlorobenzenecarboxaldehyde, 4-chloro-benzaldehyde, 4-chlorobenzenaldehyde, 4-chloro benzaldehyde, ccris 857, para-chlorobenzaldehyde, unii-e67727up9z | |
AVPYQKSLYISFPO-UHFFFAOYSA-N | |
4-chlorobenzaldehyde | |
7726 | |
140.57 | |
Crystalline Mass |
>46.0°C | |
1.1960g/mL | |
88°C | |
97.5% min. (GC) | |
ClC6H4CHO | |
1.196 | |
Solubility in water: 935mg/l (20°C) | |
C1=CC(=CC=C1C=O)Cl | |
140.57 | |
CHEBI:28105 | |
98% | |
4-Chlorobenzaldehyde |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Toxic to aquatic life with long lasting effects.
GHS P Statement
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN:
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 203-247-4
- 4-Chlorobenzaldehyde
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P310-Immediately call a POISON CENTER/doctor/.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P273-Avoid release to the environment.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.