Learn More
4-Chloropyridine hydrochloride, 97%, Thermo Scientific Chemicals
67.60 € - 189.00 €
Chemical Identifiers
CAS | 7379-35-3 |
---|---|
Molecular Formula | C5H5Cl2N |
Molecular Weight (g/mol) | 150.00 |
MDL Number | MFCD00012829 |
InChI Key | XGAFCCUNHIMIRV-UHFFFAOYSA-N |
Synonym | 4-chloropyridine hydrochloride, 4-chloropyridine hcl, 4-chloropyridinium chloride, pyridine, 4-chloro-, hydrochloride, 4-chloro-pyridine hydrochloride, ccris 1716, pyridine, 4-chloro-, hydrochloride 1:1, 4-chloropyridine, chloride, 4-chloropyridinehydrochloride, 4-chloropyrodine hydrochloride |
PubChem CID | 81852 |
IUPAC Name | 4-chloropyridine;hydrochloride |
SMILES | [H+].[Cl-].ClC1=CC=NC=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10336220
|
Thermo Scientific
110030250 |
25 g | Glass bottle |
67.60 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10530011
|
Thermo Scientific Acros
110031000 |
100 g | Plastic Bottle |
189.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7379-35-3 | |
150.00 | |
XGAFCCUNHIMIRV-UHFFFAOYSA-N | |
81852 | |
[H+].[Cl-].ClC1=CC=NC=C1 |
C5H5Cl2N | |
MFCD00012829 | |
4-chloropyridine hydrochloride, 4-chloropyridine hcl, 4-chloropyridinium chloride, pyridine, 4-chloro-, hydrochloride, 4-chloro-pyridine hydrochloride, ccris 1716, pyridine, 4-chloro-, hydrochloride 1:1, 4-chloropyridine, chloride, 4-chloropyridinehydrochloride, 4-chloropyrodine hydrochloride | |
4-chloropyridine;hydrochloride |
Specifications
7379-35-3 | |
100.0 | |
Beige-Yellow to White | |
Authentic | |
C5H5Cl2N | |
MFCD00012829 | |
4-chloropyridine hydrochloride, 4-chloropyridine hcl, 4-chloropyridinium chloride, pyridine, 4-chloro-, hydrochloride, 4-chloro-pyridine hydrochloride, ccris 1716, pyridine, 4-chloro-, hydrochloride 1:1, 4-chloropyridine, chloride, 4-chloropyridinehydrochloride, 4-chloropyrodine hydrochloride | |
XGAFCCUNHIMIRV-UHFFFAOYSA-N | |
4-chloropyridine;hydrochloride | |
81852 | |
97% | |
4-Chloropyridine hydrochloride, 97% |
96.0 | |
210.0°C | |
198°C | |
96% min. (HPLC) | |
(C5H4N)Cl | |
20, 231 | |
Solubility in water: soluble | |
[H+].[Cl-].ClC1=CC=NC=C1 | |
150.00 | |
150.01 | |
Crystalline Powder |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 230-946-1
missing translation for 'tsca' : TSCA
RUO – Research Use Only