missing translation for 'onlineSavingsMsg'
Learn More
4-Dimethylaminobenzaldehyde, 98% Product Code.: 11489223

4-Dimethylaminobenzaldehyde, 98%

Product Code. 11489223
100 g, 100g
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
This item is not returnable. View return policy

Product Code. 11489223

Brand: Thermo Scientific Alfa Aesar A11712.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 100-10-7 | C9H11NO | 149.19 g/mol

4-Dimethylaminobenzaldehyde is used to prepare colorful Schiff base adducts with amines, pyrroles and indoles. It is often used in the Ehrlich reaction, test for the presence of indoles in a sample containing alkaloids and determination of hydrazine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Dimethylaminobenzaldehyde is used to prepare colorful Schiff base adducts with amines, pyrroles and indoles. It is often used in the Ehrlich reaction, test for the presence of indoles in a sample containing alkaloids and determination of hydrazine.

Solubility
Soluble in alcohol, ether, chloroform, acetic acid. Insoluble in water.

Notes
Light sensitive. Stable at room temperature.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-10-7
Molecular Formula C9H11NO
Molecular Weight (g/mol) 149.19
MDL Number MFCD00003381
InChI Key BGNGWHSBYQYVRX-UHFFFAOYSA-N
Synonym 4-dimethylamino benzaldehyde, p-dimethylaminobenzaldehyde, ehrlich's reagent, p-formyldimethylaniline, benzaldehyde, 4-dimethylamino, p-dimethylamino benzaldehyde, p-formyl-n,n-dimethylaniline, n,n-dimethyl-p-aminobenzaldehyde, p-n,n-dimethylamino benzaldehyde, 4-dimethylaminobenzenecarbonalShow MoreShow Less
PubChem CID 7479
IUPAC Name 4-(dimethylamino)benzaldehyde
SMILES CN(C)C1=CC=C(C=O)C=C1

Specifications

Melting Point 73°C to 75°C
Boiling Point 176°C to 177°C (17 mmHg)
Flash Point 164°C (327°F)
Odor Strong
Quantity 100 g
Beilstein 606802
Sensitivity Air and light sensitive
Merck Index 14,3230
Solubility Information Soluble in alcohol,ether,chloroform,acetic acid. Insoluble in water.
Formula Weight 149.19
Percent Purity 98%
Chemical Name or Material 4-Dimethylaminobenzaldehyde
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Product Code
Lot Number10250849 Certificate TypeCertificate of Analysis Date30/08/2024 Product CodeA11712.0E,A11712.36,A11712.22
Lot Number10193612 Certificate TypeCertificate of Analysis Date05/02/2024 Product CodeA11712.0E,A11712.36,A11712.22
Lot Number10155464 Certificate TypeCertificate of Analysis Date05/02/2024 Product CodeA11712.0E,A11712.36,A11712.22
Lot Number10241388 Certificate TypeCertificate of Analysis Date21/06/2022 Product CodeA11712.0E,A11712.36,A11712.22
Lot Number10158523 Certificate TypeCertificate of Analysis Date22/11/2021 Product CodeA11712.0E,A11712.36,A11712.22
Product Identifier
  • 4-Dimethylaminobenzaldehyde
Supplemental information
  • MIXTURE LIST-Contains : 4-Dimethylaminobenzaldehyde

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Dimethylaminobenzaldehyde, 98% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.