missing translation for 'onlineSavingsMsg'
Learn More
4-Isobutyl-alpha-methylphenylacetic acid, 99%, Thermo Scientific Chemicals Product Code.: 11440067

4-Isobutyl-alpha-methylphenylacetic acid, 99%, Thermo Scientific Chemicals

Product Code. 11440067
5 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 11440067

Brand: Thermo Scientific Alfa Aesar B20989.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Ibuprofen is useful as a nonsteroidal anti-inflammatory research chemical. Ibuprofen is reported to inhibit the netrophil-dependent edema response of rabbit skin to C5a/PGE23. NSAID that inhibits cyclooxygenase-1 and -2.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ibuprofen is useful as a nonsteroidal anti-inflammatory research chemical. Ibuprofen is reported to inhibit the netrophil-dependent edema response of rabbit skin to C5a/PGE23. NSAID that inhibits cyclooxygenase-1 and -2.

Solubility
Insoluble in water. Soluble in ethanol (25 mg/ml), chloroform (1:1), ether (1:2), acetone (1:1.5), aqueous solutions of alkali hydroxides and carbonates, dichloromethane, methanol (50 mg/ml), and ethyl acetate.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15687-27-1
Molecular Formula C13H18O2
Molecular Weight (g/mol) 206.29
MDL Number MFCD00010393
InChI Key HEFNNWSXXWATRW-UHFFFAOYNA-N
Synonym ibuprofen, 2-4-isobutylphenyl propanoic acid, motrin, brufen, nurofen, advil, dolgit, liptan, nuprin, anflagen
PubChem CID 3672
ChEBI CHEBI:5855
IUPAC Name 2-[4-(2-methylpropyl)phenyl]propanoic acid
SMILES CC(C)CC1=CC=C(C=C1)C(C)C(O)=O

Specifications

Melting Point 75°C to 78°C
Boiling Point 157°C (4 mmHg)
Odor Odorless
Quantity 5 g
UN Number UN3077
Merck Index 14,4881
Solubility Information Insoluble in water. Soluble in ethanol (25mg/ml),chloroform (1:1),ether (1:2),acetone (1:1.5),aqueous solutions of alkali hydroxides and carbonates,dichloromethane,methanol (50mg/ml),and ethyl acetate.
Formula Weight 206.29
Percent Purity 99%
Chemical Name or Material 4-Isobutyl-alpha-methylphenylacetic acid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberR24J013 Certificate TypeCertificate of Analysis Date06/06/2023 Catalog NumberB20989.06
Lot Number10240898 Certificate TypeCertificate of Analysis Date30/05/2022 Catalog NumberB20989.06,B20989.03,B20989.14
Lot NumberM31I034 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberB20989.06,B20989.03,B20989.14
Lot Number10236227 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB20989.06,B20989.03,B20989.14
Lot NumberS18E032 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB20989.06,B20989.03,B20989.14
compliance-icons
Product Identifier
  • 4-Isobutyl-alpha-methylphenylacetic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : 4-Isobutyl-alpha-methylphenylacetic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Isobutyl-alpha-methylphenylacetic acid, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.