Learn More
4'-Isopropylacetophenone, 97%, Thermo Scientific Chemicals
29.35 € - 261.00 €
Chemical Identifiers
CAS | 645-13-6 |
---|---|
Molecular Formula | C11H14O |
Molecular Weight (g/mol) | 162.23 |
MDL Number | MFCD00048297 |
InChI Key | PDLCCNYKIIUWHA-UHFFFAOYSA-N |
Synonym | 4'-isopropylacetophenone, 4-isopropylacetophenone, p-isopropylacetophenone, cuminone, 1-4-isopropylphenyl ethanone, ethanone, 1-4-1-methylethyl phenyl, acetophenone, 4'-isopropyl, unii-742zkz2a6d, methyl p-isopropylphenyl ketone, 1-4-propan-2-yl phenyl ethan-1-one |
PubChem CID | 12578 |
IUPAC Name | 1-(4-propan-2-ylphenyl)ethanone |
SMILES | CC(C)C1=CC=C(C=C1)C(C)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11448977
|
Thermo Scientific Alfa Aesar
B20581.06 |
5 g |
29.35 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11458977
|
Thermo Scientific Alfa Aesar
B20581.14 |
25 g |
93.06 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11468977
|
Thermo Scientific Alfa Aesar
B20581.22 |
100 g |
261.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
4'-Isopropylacetophenone is used as perfuming agents. It is also used as flavoring and fragrance agent. Indenes are yielded from the derivatives if this compound in one step synthesis using a heterogeneous catalyst.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4′-Isopropylacetophenone is used as perfuming agents. It is also used as flavoring and fragrance agent. Indenes are yielded from the derivatives if this compound in one step synthesis using a heterogeneous catalyst.
Solubility
Soluble in alcohol. Insoluble in water.
Notes
Store at room temperature. Incompatible with strong oxidizing agents.
Chemical Identifiers
645-13-6 | |
162.23 | |
PDLCCNYKIIUWHA-UHFFFAOYSA-N | |
12578 | |
CC(C)C1=CC=C(C=C1)C(C)=O |
C11H14O | |
MFCD00048297 | |
4'-isopropylacetophenone, 4-isopropylacetophenone, p-isopropylacetophenone, cuminone, 1-4-isopropylphenyl ethanone, ethanone, 1-4-1-methylethyl phenyl, acetophenone, 4'-isopropyl, unii-742zkz2a6d, methyl p-isopropylphenyl ketone, 1-4-propan-2-yl phenyl ethan-1-one | |
1-(4-propan-2-ylphenyl)ethanone |
Specifications
645-13-6 | |
238°C | |
1.523 | |
2205694 | |
Soluble in alcohol. Insoluble in water. | |
CC(C)C1=CC=C(C=C1)C(C)=O | |
162.23 | |
162.23 | |
4'-Isopropylacetophenone |
0.97 | |
C11H14O | |
MFCD00048297 | |
4'-isopropylacetophenone, 4-isopropylacetophenone, p-isopropylacetophenone, cuminone, 1-4-isopropylphenyl ethanone, ethanone, 1-4-1-methylethyl phenyl, acetophenone, 4'-isopropyl, unii-742zkz2a6d, methyl p-isopropylphenyl ketone, 1-4-propan-2-yl phenyl ethan-1-one | |
PDLCCNYKIIUWHA-UHFFFAOYSA-N | |
1-(4-propan-2-ylphenyl)ethanone | |
12578 | |
98% |
Safety and Handling
missing translation for 'einecsNumber' : 211-433-1
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only