missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(Isopropylcarbamoyl)benzeneboronic acid, 95%, Thermo Scientific Chemicals
56.50 € - 173.00 €
Chemical Identifiers
CAS | 397843-67-3 |
---|---|
Molecular Formula | C10H14BNO3 |
Molecular Weight (g/mol) | 207.036 |
MDL Number | MFCD04039353 |
InChI Key | GBCSEYKTZAKRMT-UHFFFAOYSA-N |
Synonym | 4-isopropylcarbamoyl phenyl boronic acid, 4-n-isopropylaminocarbonyl phenylboronic acid, 4-isopropylcarbamoyl benzeneboronic acid, 4-isopropylcarbamoyl phenylboronic acid, 4-propan-2-ylcarbamoyl phenyl boronic acid, 4-n-isopropylaminocarbonyl phenyl boronic acid, n-isopropyl 4-boronobenzamide, boronic acid, 4-1-methylethyl amino carbonyl phenyl, 4-1-methylethyl amino carbonyl phenyl-boronic acid |
PubChem CID | 2773475 |
IUPAC Name | [4-(propan-2-ylcarbamoyl)phenyl]boronic acid |
SMILES | B(C1=CC=C(C=C1)C(=O)NC(C)C)(O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15460216
|
Thermo Scientific Alfa Aesar
H52925.MD |
250 mg |
56.50 €
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15439697
|
Thermo Scientific Alfa Aesar
H52925.03 |
1 g |
173.00 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
397843-67-3 | |
207.036 | |
GBCSEYKTZAKRMT-UHFFFAOYSA-N | |
2773475 | |
B(C1=CC=C(C=C1)C(=O)NC(C)C)(O)O |
C10H14BNO3 | |
MFCD04039353 | |
4-isopropylcarbamoyl phenyl boronic acid, 4-n-isopropylaminocarbonyl phenylboronic acid, 4-isopropylcarbamoyl benzeneboronic acid, 4-isopropylcarbamoyl phenylboronic acid, 4-propan-2-ylcarbamoyl phenyl boronic acid, 4-n-isopropylaminocarbonyl phenyl boronic acid, n-isopropyl 4-boronobenzamide, boronic acid, 4-1-methylethyl amino carbonyl phenyl, 4-1-methylethyl amino carbonyl phenyl-boronic acid | |
[4-(propan-2-ylcarbamoyl)phenyl]boronic acid |
Specifications
397843-67-3 | |
C10H14BNO3 | |
4-isopropylcarbamoyl phenyl boronic acid, 4-n-isopropylaminocarbonyl phenylboronic acid, 4-isopropylcarbamoyl benzeneboronic acid, 4-isopropylcarbamoyl phenylboronic acid, 4-propan-2-ylcarbamoyl phenyl boronic acid, 4-n-isopropylaminocarbonyl phenyl boronic acid, n-isopropyl 4-boronobenzamide, boronic acid, 4-1-methylethyl amino carbonyl phenyl, 4-1-methylethyl amino carbonyl phenyl-boronic acid | |
B(C1=CC=C(C=C1)C(=O)NC(C)C)(O)O | |
207.036 | |
207.04 | |
4-(Isopropylcarbamoyl)benzeneboronic acid |
162°C to 167°C | |
MFCD04039353 | |
GBCSEYKTZAKRMT-UHFFFAOYSA-N | |
[4-(propan-2-ylcarbamoyl)phenyl]boronic acid | |
2773475 | |
95% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only