Learn More
4-Methoxybenzenediazonium tetrafluoroborate, 98%, Thermo Scientific Chemicals
34.07 € - 460.29 €
Chemical Identifiers
CAS | 459-64-3 |
---|---|
Molecular Formula | C7H7BF4N2O |
Molecular Weight (g/mol) | 221.95 |
MDL Number | MFCD00011897 |
InChI Key | CACFTKIREZJSIG-UHFFFAOYSA-J |
Synonym | 4-methoxybenzenediazonium tetrafluoroborate, 4-methoxybenzenediazoniumfluoroborate, 4-methoxybenzene diazonium fluoroborate, acmc-1aeuk, ksc236e8f, 4-methoxyphenyldiazonium tetrafluoroborate, p-methoxybenzenediazonium tetrafluoroborate, 4-methoxybenzene-1-diazonium; tetrafluoroboranuideShow More |
PubChem CID | 517233 |
SMILES | [B+3].[F-].[F-].[F-].[F-].COC1=CC=C(C=C1)[N+]#N |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15432188
|
Thermo Scientific Alfa Aesar
H55827.03 |
1 g |
34.07 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15442188
|
Thermo Scientific Alfa Aesar
H55827.06 |
5 g |
109.55 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15452188
|
Thermo Scientific Alfa Aesar
H55827.14 |
25 g |
460.29 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
4-Methoxybenzenediazonium tetrafluoroborate has been used in the preparation of azo coupled cyclic β-enaminones.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Methoxybenzenediazonium tetrafluoroborate has been used in the preparation of azo coupled cyclic β-enaminones.
Solubility
Decompose in water
Notes
Store at -20°C. Light sensitive.
Chemical Identifiers
459-64-3 | |
221.95 | |
CACFTKIREZJSIG-UHFFFAOYSA-J | |
517233 |
C7H7BF4N2O | |
MFCD00011897 | |
4-methoxybenzenediazonium tetrafluoroborate, 4-methoxybenzenediazoniumfluoroborate, 4-methoxybenzene diazonium fluoroborate, acmc-1aeuk, ksc236e8f, 4-methoxyphenyldiazonium tetrafluoroborate, p-methoxybenzenediazonium tetrafluoroborate, 4-methoxybenzene-1-diazonium; tetrafluoroboranuideShow More | |
[B+3].[F-].[F-].[F-].[F-].COC1=CC=C(C=C1)[N+]#N |
Specifications
459-64-3 | |
C7H7BF4N2O | |
UN3261 | |
Decompose in water | |
[B+3].[F-].[F-].[F-].[F-].COC1=CC=C(C=C1)[N+]#N | |
221.95 | |
221.95 | |
4-Methoxybenzenediazonium tetrafluoroborate |
142°C to 144°C | |
MFCD00011897 | |
4-methoxybenzenediazonium tetrafluoroborate, 4-methoxybenzenediazoniumfluoroborate, 4-methoxybenzene diazonium fluoroborate, acmc-1aeuk, ksc236e8f, 4-methoxyphenyldiazonium tetrafluoroborate, p-methoxybenzenediazonium tetrafluoroborate, 4-methoxybenzene-1-diazonium; tetrafluoroboranuide | |
CACFTKIREZJSIG-UHFFFAOYSA-J | |
4-methoxybenzenediazonium;tetrafluoroborate | |
517233 | |
98% |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302+H312+H332-H314
missing translation for 'dotInformation' : Hazard Class: 8; Packaging Group: II
missing translation for 'einecsNumber' : 207-296-2
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.