missing translation for 'onlineSavingsMsg'
Learn More
4-Methoxybenzoyl chloride, 97%, Thermo Scientific Chemicals Product Code.: 11377906

4-Methoxybenzoyl chloride, 97%, Thermo Scientific Chemicals

Product Code. 11377906
25 g
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy

Product Code. 11377906

Brand: Thermo Scientific Alfa Aesar L13120.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

4-Methoxybenzoyl chloride is used in the synthesis of stilbene and dihydrostilbene derivatives as potential anti-cancer agents. It is also used in the synthesis of coumarin dimers with potential HIV-1 activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methoxybenzoyl chloride is used in the synthesis of stilbene and dihydrostilbene derivatives as potential anti-cancer agents. It is also used in the synthesis of coumarin dimers with potential HIV-1 activity.

Solubility
It reacts in water.

Notes
Moisture sensitive. Incompatible with oxidizing agents and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-07-2
Molecular Formula C8H7ClO2
Molecular Weight (g/mol) 170.59
MDL Number MFCD00000687
InChI Key MXMOTZIXVICDSD-UHFFFAOYSA-N
Synonym p-anisoyl chloride, benzoyl chloride, 4-methoxy, anisoyl chloride, 4-anisoyl chloride, p-methoxybenzoyl chloride, 4-methoxybenzoylchloride, 4-methoxybenzoic acid chloride, benzoyl chloride, p-methoxy, p-methoxybenzoic acid chloride, 4-methoxy-benzoyl chlorideShow MoreShow Less
PubChem CID 7477
IUPAC Name 4-methoxybenzoyl chloride
SMILES COC1=CC=C(C=C1)C(Cl)=O

Specifications

Melting Point 22°C
Density 1.26
Boiling Point 145°C (14 mmHg)
Flash Point 87°C (188°F)
Refractive Index 1.581
Quantity 25 g
UN Number UN1729
Beilstein 471918
Sensitivity Moisture sensitive
Merck Index 14,673
Solubility Information It reacts in water.
Formula Weight 170.6
Percent Purity 97%
Chemical Name or Material 4-Methoxybenzoyl chloride
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10244615 Certificate TypeCertificate of Analysis Date16/11/2022 Catalog NumberL13120.22,L13120.14
Lot Number5001P31R Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL13120.22,L13120.14
Lot Number10205398 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL13120.22,L13120.14
Lot Number10224168 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL13120.22,L13120.14
Lot NumberL17T041 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL13120.22,L13120.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Methoxybenzoyl chloride, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.