Learn More
4-Methoxybenzoyl chloride, 97%, Thermo Scientific Chemicals
34.17 € - 84.22 €
Chemical Identifiers
CAS | 100-07-2 |
---|---|
Molecular Formula | C8H7ClO2 |
Molecular Weight (g/mol) | 170.59 |
MDL Number | MFCD00000687 |
InChI Key | MXMOTZIXVICDSD-UHFFFAOYSA-N |
Synonym | p-anisoyl chloride, benzoyl chloride, 4-methoxy, anisoyl chloride, 4-anisoyl chloride, p-methoxybenzoyl chloride, 4-methoxybenzoylchloride, 4-methoxybenzoic acid chloride, benzoyl chloride, p-methoxy, p-methoxybenzoic acid chloride, 4-methoxy-benzoyl chloride |
PubChem CID | 7477 |
IUPAC Name | 4-methoxybenzoyl chloride |
SMILES | COC1=CC=C(C=C1)C(Cl)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11377906
|
Thermo Scientific Alfa Aesar
L13120.14 |
25 g |
34.17 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11387906
|
Thermo Scientific Alfa Aesar
L13120.22 |
100 g |
84.22 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
4-Methoxybenzoyl chloride is used in the synthesis of stilbene and dihydrostilbene derivatives as potential anti-cancer agents. It is also used in the synthesis of coumarin dimers with potential HIV-1 activity.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Methoxybenzoyl chloride is used in the synthesis of stilbene and dihydrostilbene derivatives as potential anti-cancer agents. It is also used in the synthesis of coumarin dimers with potential HIV-1 activity.
Solubility
It reacts in water.
Notes
Moisture sensitive. Incompatible with oxidizing agents and bases.
Chemical Identifiers
100-07-2 | |
170.59 | |
MXMOTZIXVICDSD-UHFFFAOYSA-N | |
7477 | |
COC1=CC=C(C=C1)C(Cl)=O |
C8H7ClO2 | |
MFCD00000687 | |
p-anisoyl chloride, benzoyl chloride, 4-methoxy, anisoyl chloride, 4-anisoyl chloride, p-methoxybenzoyl chloride, 4-methoxybenzoylchloride, 4-methoxybenzoic acid chloride, benzoyl chloride, p-methoxy, p-methoxybenzoic acid chloride, 4-methoxy-benzoyl chloride | |
4-methoxybenzoyl chloride |
Specifications
100-07-2 | |
1.26 | |
87°C (188°F) | |
1.581 | |
UN1729 | |
Moisture sensitive | |
p-anisoyl chloride, benzoyl chloride, 4-methoxy, anisoyl chloride, 4-anisoyl chloride, p-methoxybenzoyl chloride, 4-methoxybenzoylchloride, 4-methoxybenzoic acid chloride, benzoyl chloride, p-methoxy, p-methoxybenzoic acid chloride, 4-methoxy-benzoyl chloride | |
MXMOTZIXVICDSD-UHFFFAOYSA-N | |
4-methoxybenzoyl chloride | |
7477 | |
97% |
22°C | |
145°C (14 mmHg) | |
C8H7ClO2 | |
MFCD00000687 | |
471918 | |
14,673 | |
It reacts in water. | |
COC1=CC=C(C=C1)C(Cl)=O | |
170.59 | |
170.6 | |
4-Methoxybenzoyl chloride |
Safety and Handling
GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
P210-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H227-H314
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: ANISOYL CHLORIDE
missing translation for 'einecsNumber' : 202-816-4
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.