missing translation for 'onlineSavingsMsg'
Learn More

4-Methylphenoxyacetic acid, 98%, Thermo Scientific Chemicals

30.10 € - 430.00 €

Chemical Identifiers

CAS 940-64-7
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.176
MDL Number MFCD00014365
InChI Key SFTDDFBJWUWKMN-UHFFFAOYSA-N
Synonym 4-methylphenoxy acetic acid, 4-methylphenoxyacetic acid, 2-4-methylphenoxy acetic acid, p-methylphenoxyacetic acid, unii-wwy23322ik, acetic acid, 4-methylphenoxy, 2-p-tolyloxy acetic acid, p-toloxyacetic acid, acmc-20apmz, p-tolyloxy-acetic acid
PubChem CID 70329
IUPAC Name 2-(4-methylphenoxy)acetic acid
SMILES CC1=CC=C(C=C1)OCC(=O)O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11440753
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A13230.09
10 g
30.10 €
10g
Please to purchase this item. Need a web account? Register with us today!
11450753
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A13230.18
50 g
112.00 €
50g
Please to purchase this item. Need a web account? Register with us today!
11460753
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A13230.30
250 g
430.00 €
250g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

940-64-7
166.176
SFTDDFBJWUWKMN-UHFFFAOYSA-N
70329
CC1=CC=C(C=C1)OCC(=O)O
C9H10O3
MFCD00014365
4-methylphenoxy acetic acid, 4-methylphenoxyacetic acid, 2-4-methylphenoxy acetic acid, p-methylphenoxyacetic acid, unii-wwy23322ik, acetic acid, 4-methylphenoxy, 2-p-tolyloxy acetic acid, p-toloxyacetic acid, acmc-20apmz, p-tolyloxy-acetic acid
2-(4-methylphenoxy)acetic acid

Specifications

940-64-7
C9H10O3
2047591
SFTDDFBJWUWKMN-UHFFFAOYSA-N
2-(4-methylphenoxy)acetic acid
70329
98%
137°C to 142°C
MFCD00014365
4-methylphenoxy acetic acid, 4-methylphenoxyacetic acid, 2-4-methylphenoxy acetic acid, p-methylphenoxyacetic acid, unii-wwy23322ik, acetic acid, 4-methylphenoxy, 2-p-tolyloxy acetic acid, p-toloxyacetic acid, acmc-20apmz, p-tolyloxy-acetic acid
CC1=CC=C(C=C1)OCC(=O)O
166.176
166.18
4-Methylphenoxyacetic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

missing translation for 'einecsNumber' : 213-374-7

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

Product Identifier
  • (4-Methylphenoxy)acetic acid

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Supplemental information
  • MIXTURE LIST-Contains : (4-Methylphenoxy)acetic acid

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only