missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Methylthiazole-5-carboxylic acid, 97%, Thermo Scientific™
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Specifications
Specifications
Melting Point | 287.0°C |
Assay Percent Range | 96% min. (HPLC) |
Packaging | Glass bottle |
Quantity | 25 g |
Formula Weight | 143.17 |
Percent Purity | 97% |
Chemical Name or Material | 4-Methylthiazole-5-carboxylic acid |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 7
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
1 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0334782 | Certificate of Analysis | 25/08/2023 | 396581000,396580250 |
Safety and Handling

Product Identifier
- 4-Methylthiazole-5-carboxylic acid
- Warning
- Respiratory sensitiser Category 1
- H317-May cause an allergic skin reaction.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
4-Methylthiazole-5-carboxylic acid, 97%, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction