missing translation for 'onlineSavingsMsg'
Learn More

4-n-Butylphenylboronic acid, 95%, Thermo Scientific™

35.41 € - 116.00 €

Chemical Identifiers

CAS 145240-28-4
Molecular Formula C10H15BO2
Molecular Weight (g/mol) 178.04
InChI Key UGZUUTHZEATQAM-UHFFFAOYSA-N
Synonym 4-butylphenyl boronic acid, 4-n-butylphenylboronic acid, 4-butylphenylboronicacid, 4-butylbenzeneboronic acid, 4-n-butylbenzeneboronic acid, 4-butylphenyl boranediol, boronic acid, 4-butylphenyl, pubchem18523, acmc-209cue, 4-butylphenylboranic acid
PubChem CID 4100860
IUPAC Name (4-butylphenyl)boronic acid
SMILES B(C1=CC=C(C=C1)CCCC)(O)O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11945871
View Documents
Thermo Scientific Acros
393870010
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11935871
View Documents
Thermo Scientific Acros
393870050
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Specifications

Chemical Identifiers

145240-28-4
178.04
4-butylphenyl boronic acid, 4-n-butylphenylboronic acid, 4-butylphenylboronicacid, 4-butylbenzeneboronic acid, 4-n-butylbenzeneboronic acid, 4-butylphenyl boranediol, boronic acid, 4-butylphenyl, pubchem18523, acmc-209cue, 4-butylphenylboranic acid
(4-butylphenyl)boronic acid
C10H15BO2
UGZUUTHZEATQAM-UHFFFAOYSA-N
4100860
B(C1=CC=C(C=C1)CCCC)(O)O

Specifications

145240-28-4
94% min. (HPLC)
C10H15BO2
4-butylphenyl boronic acid, 4-n-butylphenylboronic acid, 4-butylphenylboronicacid, 4-butylbenzeneboronic acid, 4-n-butylbenzeneboronic acid, 4-butylphenyl boranediol, boronic acid, 4-butylphenyl, pubchem18523, acmc-209cue, 4-butylphenylboranic acid
B(C1=CC=C(C=C1)CCCC)(O)O
178.04
178.04
4-n-Butylphenylboronic acid
91.0°C to 97.0°C
Glass Bottle
CH3(CH2)3C6H4B(OH)2
UGZUUTHZEATQAM-UHFFFAOYSA-N
(4-butylphenyl)boronic acid
4100860
95%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
If eye irritation persists: Get medical advice/attention.
Call a POISON CENTER or doctor/phys

GHS Signal Word: Warning

Product Identifier
  • 4-n-Butylphenylboronic acid

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only