Learn More
4-Nitrobenzoyl chloride, 98%, Thermo Scientific Chemicals
24.92 € - 119.60 €
Chemical Identifiers
CAS | 122-04-3 |
---|---|
Molecular Formula | C7H4ClNO3 |
Molecular Weight (g/mol) | 185.56 |
MDL Number | MFCD00007345 |
InChI Key | SKDHHIUENRGTHK-UHFFFAOYSA-N |
Synonym | p-nitrobenzoyl chloride, benzoyl chloride, 4-nitro, 4-nitrobenzoic acid chloride, p-nitrobenzoic acid chloride, benzoyl chloride, p-nitro, 4-nitro-benzoyl chloride, unii-x3h8pw2gc4, ccris 3136, 4-nitrobenzoylchloride, 4-nitro benzoyl chloride |
PubChem CID | 8502 |
IUPAC Name | 4-nitrobenzoyl chloride |
SMILES | [O-][N+](=O)C1=CC=C(C=C1)C(Cl)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10337610
|
Thermo Scientific Acros
150430050 |
5 g | Glass bottle |
24.92 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10497920
|
Thermo Scientific Acros
150431000 |
100 g | Glass bottle |
33.37 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10041570
|
Thermo Scientific Acros
150432500 |
250 g | Glass bottle |
64.92 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10503701
|
Thermo Scientific Acros
150435000 |
500 g | Glass bottle |
119.60 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Currently Unavailable: We have no information when this product will be available
View alternative products
|
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
122-04-3 | |
185.56 | |
SKDHHIUENRGTHK-UHFFFAOYSA-N | |
8502 | |
[O-][N+](=O)C1=CC=C(C=C1)C(Cl)=O |
C7H4ClNO3 | |
MFCD00007345 | |
p-nitrobenzoyl chloride, benzoyl chloride, 4-nitro, 4-nitrobenzoic acid chloride, p-nitrobenzoic acid chloride, benzoyl chloride, p-nitro, 4-nitro-benzoyl chloride, unii-x3h8pw2gc4, ccris 3136, 4-nitrobenzoylchloride, 4-nitro benzoyl chloride | |
4-nitrobenzoyl chloride |
Specifications
122-04-3 | |
100.0 | |
Yellow | |
102°C | |
98% | |
O2NC6H4COCl | |
09, 394 | |
15, 6677 | |
Solubility in water: decomposes in water. Other solubilities: decomposes in alcohol,soluble in diethyl ether | |
[O-][N+](=O)C1=CC=C(C=C1)C(Cl)=O | |
185.56 | |
185.57 | |
Crystals or Flakes |
97.5 | |
70°C to 74°C | |
202°C to 205°C (105.0 mmHg) | |
Authentic | |
C7H4ClNO3 | |
MFCD00007345 | |
01,736 | |
p-nitrobenzoyl chloride, benzoyl chloride, 4-nitro, 4-nitrobenzoic acid chloride, p-nitrobenzoic acid chloride, benzoyl chloride, p-nitro, 4-nitro-benzoyl chloride, unii-x3h8pw2gc4, ccris 3136, 4-nitrobenzoylchloride, 4-nitro benzoyl chloride | |
SKDHHIUENRGTHK-UHFFFAOYSA-N | |
4-nitrobenzoyl chloride | |
8502 | |
98% | |
4-Nitrobenzoyl chloride, 98% |
Safety and Handling
GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Contact with water liberates toxic gas.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 204-517-4
missing translation for 'rtecsNumber' : DM6651000
missing translation for 'tsca' : TSCA
- 4-Nitrobenzoyl chloride
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- EUH029-Contact with water liberates toxic gas.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P402+P404-Store in a dry place. Store in a closed container.
RUO – Research Use Only