missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrophenethyl alcohol, 99%, Thermo Scientific Chemicals

84.02 € - 298.00 €

Chemical Identifiers

CAS 100-27-6
Molecular Formula C8H9NO3
Molecular Weight (g/mol) 167.16
InChI Key IKMXRUOZUUKSON-UHFFFAOYSA-N
Synonym 4-nitrophenethyl alcohol, 2-4-nitrophenyl ethanol, 4-nitrobenzeneethanol, 2-p-nitrophenyl ethanol, benzeneethanol, 4-nitro, p-nitrophenethyl alcohol, 2-4-nitrophenyl ethan-1-ol, phenethyl alcohol, p-nitro, ccris 6079, 4-nitrophenethylalcohol
PubChem CID 7494
IUPAC Name 2-(4-nitrophenyl)ethanol
SMILES C1=CC(=CC=C1CCO)[N+](=O)[O-]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15505056
View Documents Promotion Details
Thermo Scientific Acros
179610100
10 g
84.02 €
10g
Please to purchase this item. Need a web account? Register with us today!
15515056
View Documents Promotion Details
Thermo Scientific Acros
179610500
50 g
298.00 €
50g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

100-27-6
167.16
4-nitrophenethyl alcohol, 2-4-nitrophenyl ethanol, 4-nitrobenzeneethanol, 2-p-nitrophenyl ethanol, benzeneethanol, 4-nitro, p-nitrophenethyl alcohol, 2-4-nitrophenyl ethan-1-ol, phenethyl alcohol, p-nitro, ccris 6079, 4-nitrophenethylalcohol
2-(4-nitrophenyl)ethanol
C8H9NO3
IKMXRUOZUUKSON-UHFFFAOYSA-N
7494
C1=CC(=CC=C1CCO)[N+](=O)[O-]

Specifications

100-27-6
148.0°C to 149.0°C (2.0 Torr)
98.5% min. (HPLC)
O2NC6H4CH2CH2OH
4-nitrophenethyl alcohol, 2-4-nitrophenyl ethanol, 4-nitrobenzeneethanol, 2-p-nitrophenyl ethanol, benzeneethanol, 4-nitro, p-nitrophenethyl alcohol, 2-4-nitrophenyl ethan-1-ol, phenethyl alcohol, p-nitro, ccris 6079, 4-nitrophenethylalcohol
C1=CC(=CC=C1CCO)[N+](=O)[O-]
167.16
167.16
4-Nitrophenethyl alcohol
56.0°C to 57.0°C
Authentic
C8H9NO3
06,II,238
IKMXRUOZUUKSON-UHFFFAOYSA-N
2-(4-nitrophenyl)ethanol
7494
99%
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 202-835-8

Product Identifier
  • 4-Nitrophenethyl alcohol, 99%

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only