missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrophenol, 99%, Thermo Scientific Chemicals

32.16 € - 392.96 €

Chemical Identifiers

CAS 100-02-7
Molecular Formula C6H5NO3
Molecular Weight (g/mol) 139.11
MDL Number MFCD00007331
InChI Key BTJIUGUIPKRLHP-UHFFFAOYSA-N
Synonym p-nitrophenol, phenol, 4-nitro, paranitrophenol, niphen, 4-hydroxynitrobenzene, p-hydroxynitrobenzene, phenol, p-nitro, mononitrophenol, paranitrofenol, paranitrofenolo
PubChem CID 980
ChEBI CHEBI:16836
IUPAC Name 4-nitrophenol
SMILES OC1=CC=C(C=C1)[N+]([O-])=O
View More Specs

Products 5
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10103051
View Documents
Thermo Scientific Acros
157050050
5 g Glass Bottle
32.16 €
5g
Please to purchase this item. Need a web account? Register with us today!
10337570
View Documents
Thermo Scientific Acros
157051000
100 g Plastic bottle
41.51 €
100g
Please to purchase this item. Need a web account? Register with us today!
10012303
View Documents
Thermo Scientific Acros
157052500
250 g Plastic bottle
53.16 €
250g
Please to purchase this item. Need a web account? Register with us today!
10675782
View Documents
Thermo Scientific Acros
157050010
1 kg Plastic bottle
107.54 €
1kg
Please to purchase this item. Need a web account? Register with us today!
11934691
View Documents
Thermo Scientific Acros
157050051
5 kg Plastic drum
392.96 €
5kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
Specifications

Chemical Identifiers

100-02-7
139.11
BTJIUGUIPKRLHP-UHFFFAOYSA-N
980
4-nitrophenol
C6H5NO3
MFCD00007331
p-nitrophenol, phenol, 4-nitro, paranitrophenol, niphen, 4-hydroxynitrobenzene, p-hydroxynitrobenzene, phenol, p-nitro, mononitrophenol, paranitrofenol, paranitrofenolo
CHEBI:16836
OC1=CC=C(C=C1)[N+]([O-])=O

Specifications

100-02-7
Yellow to Brown
169°C
98.5% min. (GC)
O2NC6H4OH
06, 226
15, 6708
Solubility in water: 1.6g/100mL (25 c). Other solubilities: soluble in alcohol,ether,benzene,chloroform and,toluene,pyridine,diethyl ether,also soluble in solns of fixed alkali hydroxides,and carbonates
OC1=CC=C(C=C1)[N+]([O-])=O
139.11
CHEBI:16836
139.11
Crystalline Powder, Crystals or Chunks
>112.0°C
279.0°C
Authentic
C6H5NO3
MFCD00007331
01,743; 02,297
p-nitrophenol, phenol, 4-nitro, paranitrophenol, niphen, 4-hydroxynitrobenzene, p-hydroxynitrobenzene, phenol, p-nitro, mononitrophenol, paranitrofenol, paranitrofenolo
BTJIUGUIPKRLHP-UHFFFAOYSA-N
4-nitrophenol
980
2.56 mPa.s (121°C)
99%
4-Nitrophenol
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause damage to organs through prolonged or repeated exposure.
Harmful if inhaled.
Harmful in contact with skin.
Toxic if swallowed.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 202-811-7

Product Identifier
  • 4-Nitrophenol

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Specific target organ toxicity after repeated exposure Category 2

Hazard Statement
  • H373-May cause damage to organs.
  • H332-Harmful if inhaled.
  • H312-Harmful in contact with skin.
  • H301-Toxic if swallowed.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only