missing translation for 'onlineSavingsMsg'
Learn More

Globe Scientific 4 oz. Specimen Containers

With EZ-Turn quarter turning screw cap designed to assist in prevention of carpal tunnel syndrome

Brand:  Globe Scientific 86775

 View more versions of this product

Product Code. 11592405

  • 132.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Polypropylene containers with polyethylene caps
  • Used for collecting urine sample
  • Graduated to 100mL with frosted writing area
  • Comes with trilingual patient ID label
  • Sterile

Specifications

Specifications

Manual
Yes
4 oz.
Green
HVP1-1
Sterile
Screw Cap
2 Ports
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.