missing translation for 'onlineSavingsMsg'
Learn More

Globe Scientific 4 oz. Specimen Containers

With EZ-Turn quarter turning screw cap designed to assist in prevention of carpal tunnel syndrome

132.00 €

Specifications

Graduated Yes
Closure Type Screw Cap
View More Specs

Products 1
Product Code Brand Color Sterility Price Quantity & Availability  
Product Code Brand Color Sterility Price Quantity & Availability  
11592405
View Documents
Globe Scientific
86775
Green Sterile
132.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
Description

Description

 

  • Polypropylene containers with polyethylene caps
  • Used for collecting urine sample
  • Graduated to 100mL with frosted writing area
  • Comes with trilingual patient ID label
  • Some units are sterile

 

Specifications

Specifications

Yes
Screw Cap
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.