Learn More
4-Pentenoic acid, 99%, Thermo Scientific Chemicals
41.00 € - 355.00 €
Chemical Identifiers
CAS | 591-80-0 |
---|---|
Molecular Formula | C5H8O2 |
Molecular Weight (g/mol) | 100.12 |
MDL Number | MFCD00004408 |
InChI Key | HVAMZGADVCBITI-UHFFFAOYSA-N |
Synonym | 4-pentenoic acid, allylacetic acid, allyl acetic acid, 4 pa, 3-vinylpropionic acid, delta 4-pentenoic acid, unii-d4s77y29fb, fema no. 2843, .delta.4-pentenoic acid, wln: qv3u1 |
PubChem CID | 61138 |
ChEBI | CHEBI:35936 |
IUPAC Name | pent-4-enoic acid |
SMILES | OC(=O)CCC=C |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10466383
|
Thermo Scientific Acros
269020050 |
5 g | Glass bottle |
41.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10731521
|
Thermo Scientific Acros
269020250 |
25 g | Glass bottle |
116.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11974961
|
Thermo Scientific Acros
269021000 |
100 g | Glass bottle |
355.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
591-80-0 | |
100.12 | |
HVAMZGADVCBITI-UHFFFAOYSA-N | |
61138 | |
pent-4-enoic acid |
C5H8O2 | |
MFCD00004408 | |
4-pentenoic acid, allylacetic acid, allyl acetic acid, 4 pa, 3-vinylpropionic acid, delta 4-pentenoic acid, unii-d4s77y29fb, fema no. 2843, .delta.4-pentenoic acid, wln: qv3u1 | |
CHEBI:35936 | |
OC(=O)CCC=C |
Specifications
591-80-0 | |
Colorless to Yellow | |
83.0°C to 84.0°C (12.0 mmHg) | |
Authentic | |
C5H8O2 | |
1.4273 to 1.4303 | |
02,425 | |
4-pentenoic acid, allylacetic acid, allyl acetic acid, 4 pa, 3-vinylpropionic acid, delta 4-pentenoic acid, unii-d4s77y29fb, fema no. 2843, .delta.4-pentenoic acid, wln: qv3u1 | |
HVAMZGADVCBITI-UHFFFAOYSA-N | |
pent-4-enoic acid | |
61138 | |
100.2 | |
Liquid |
-22.5°C | |
0.9750g/mL | |
89°C | |
98.5% min. (GC) | |
H2C=CHCH2CH2CO2H | |
MFCD00004408 | |
0.975 | |
Solubility in water: insoluble | |
OC(=O)CCC=C | |
100.12 | |
CHEBI:35936 | |
99% | |
4-Pentenoic acid |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 209-732-7
RUO – Research Use Only