missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-(piperazine-1-carbonyl)phenylboronic acid, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Specifications
Specifications
CAS | 864754-07-4 |
CAS Min % | 97.0 |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Molecular Formula | C17H25BN2O3 |
MDL Number | MFCD06657704 |
Quantity | 1 g |
Synonym | 4-piperazine-1-carbonyl phenylboronic acid, pinacol ester, piperazin-1-yl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methanone, 4-piperazine-1-carbonyl phenylboronic acid pinacol ester, piperazin-1-yl-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-phenyl-methanone, 1-4-tetramethyl-1,3,2-dioxaborolan-2-yl benzoyl piperazine, 1-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzoyl piperazine, amtb003, abbypharma ap-13-12359, methanone,1-piperazinyl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl |
InChI Key | OBWNKFAHYIGNHE-UHFFFAOYSA-N |
SMILES | B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)C(=O)N3CCNCC3 |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 9
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
4-(piperazine-1-carbonyl)phenylboronic acid, 97%, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction