missing translation for 'onlineSavingsMsg'
Learn More
4-[(tert-Butoxycarbonylamino)methyl]phenylboronic acid pinacol ester, 97%, Thermo Scientific Chemicals Product Code.: 11916061

4-[(tert-Butoxycarbonylamino)methyl]phenylboronic acid pinacol ester, 97%, Thermo Scientific Chemicals

Product Code. 11916061
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11916061

Brand: Thermo Scientific Acros 444640010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 330794-35-9
Molecular Formula C18H28BNO4
Molecular Weight (g/mol) 333.24
MDL Number MFCD06409942
InChI Key CUDCEJRRWNIPDL-UHFFFAOYSA-N
Synonym 4-n-boc-aminomethyl phenylboronic acid pinacol ester, tert-butyl 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzylcarbamate, 4-n-boc-amino methyl phenylboronic acid pinacol ester, 4-tert-butoxycarbonylaminomethyl phenylboronic acid, pinacol ester, tert-butyl n-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl methyl carbamate, 4-tert-butoxycarbonylaminomethyl phenylboronic acid pinacol ester, 4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-benzyl-carbamic acid tert-butyl ester, pubchem5247, 4-n-boc-aminomethyl phenylboronic acid,pinacol esterShow MoreShow Less
PubChem CID 3415442
IUPAC Name tert-butyl N-[[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl]carbamate
SMILES CC(C)(C)OC(=O)NCC1=CC=C(C=C1)B1OC(C)(C)C(C)(C)O1

Specifications

Melting Point 68.0°C to 69.0°C
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 333.24
Percent Purity 97%
Chemical Name or Material 4-[(tert-Butoxycarbonylamino)methyl]phenylboronic acid pinacol ester

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Catalog Number
Lot Number345263 Certificate TypeCertificate of Analysis Date20/02/2024 Catalog Number444640010,444640050
Lot Number344074 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number444640010,444640050
compliance-icons
Product Identifier
  • 4-¢(tert-Butoxycarbonylamino)methyl!phenylboronic acid pinacol ester
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-[(tert-Butoxycarbonylamino)methyl]phenylboronic acid pinacol ester, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.