Learn More
Fisherbrand™ 4-Way Clinical Rack
The 4-Way Clinical Tube Rack is the only tube rack specifically designed to hold round, flat and conical blood collection and culture tubes. It holds multiple tube sizes without multiple racks.
104.00 € - 116.00 €
Specifications
Product Type Blood Tube Rack
Includes Rack
Cell Dividers No
Clean Claims Cleaned with mild detergents and autoclavable
Closure Included No
View More Specs
Products 5
Product Code Brand Color Price Quantity & Availability  
Product Code Brand Color Price Quantity & Availability  
16340402
View Documents
Fisherbrand™
120683F
Blue, Purple, Green, Red, Natural
116.00 €
Pack of 5
Estimated Shipment: 27-05-2025
to see stock.
Add to basket
16350402
View Documents
Fisherbrand™
120684F
Blue
116.00 €
Pack of 5
In Stock
to see stock.
Edge
Add to basket
16360402
View Documents
Fisherbrand™
120685F
Purple
116.00 €
Pack of 5
In Stock
to see stock.
Edge
Add to basket
16370402
View Documents
Fisherbrand™
120686F
Green
104.00 €
Pack of 5
Estimated Shipment: 13-05-2025
to see stock.
Add to basket
16380402
View Documents
Fisherbrand™
120687F
Natural
116.00 €
Pack of 5
Estimated Shipment: 27-05-2025
to see stock.
Add to basket
Description
Description
  • Accommodates all brands of 13 and 16 mm round, flat and conical bottom blood collection and culture tubes.
  • Only 4-way rack available with a six way connection.
  • Link different or the same well sizes, end to end or side by side, in any orientation.
  • Customize the rack to fit the number of wells required, infinite units can be linked together or use one standalone unit.
  • Uniformly spaced wells help eliminate pipetting errors which enables more accurate results.
  • Molded alphanumeric identification for quick location and retrieval of tubes.
  • Robust, made from polypropylene that is chemical resistant and easy to clean.
  • Racks can be used in water baths and placed in freezers.
  • Assorted color pack available for color coding options.
  • Patent Pending
  • Autoclavable
Specifications
Show More
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Fisherbrand™ 4-Way Clinical Rack