Learn More
RETSCH 400 dia. x 65 mmH Stainless Steel Test Sieve, ASTM Certified, Millimeter Pore Sizes
Brand:  RETSCH 60.167.037500
 View more versions of this product
Product Code. 10228501
  • 482.00 € / Each
Estimated Shipment: 21-03-2025
to see stock.
Explore more special offers
Add to basket
Specifications
1Each
Test Sieve
Stainless Steel
65mm
400 mm
Stainless Steel
37.50 mm
ASTM E 11
Shaker AS 450
For separation, fractioning, particle size determination
Stainless Steel