Learn More
RETSCH 400 dia. x 65 mmH Stainless Steel Test Sieve, ASTM Certified, Millimeter Pore Sizes
Brand:  RETSCH 60.167.002000
 View more versions of this product
Product Code. 10438731
  • 304.00 € / Each
Estimated Shipment: 28-02-2025
to see stock.
Explore more special offers
Add to basket
Specifications
1Each
Test Sieve
Stainless Steel
65mm
400 mm
Stainless Steel
2 mm
ASTM E 11
Shaker AS 450
For separation, fractioning, particle size determination
Stainless Steel