missing translation for 'onlineSavingsMsg'
Learn More
(4R,6R)-tert-Butyl-6-cyanomethyl-2,2-dimethyl-1,3-dioxane-4-acetate, 97%, Thermo Scientific™ Product Code.: 10590522

(4R,6R)-tert-Butyl-6-cyanomethyl-2,2-dimethyl-1,3-dioxane-4-acetate, 97%, Thermo Scientific™

Product Code. 10590522
1 g, Glass bottle
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10590522

Brand: Thermo Scientific Acros 357990010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 125971-94-0
CAS Min % 96.0
CAS Max % 100.0
Melting Point 65.0°C to 70.0°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Molecular Formula C14H23NO4
MDL Number MFCD03093958
Quantity 1 g
Synonym 4r,6r-tert-butyl-6-cyanomethyl-2,2-dimethyl-1,3-dioxane-4-acetate, tert-butyl 4r,6r-6-cyanomethyl-2,2-dimethyl-1,3-dioxane-4-acetate, tert-butyl 2-4r,6r-6-cyanomethyl-2,2-dimethyl-1,3-dioxan-4-yl acetate, 4r,6r-t-butyl-6-cyanomethyl-2,2-dimethyl-1,3-dioxane-4-acetate, 4r-cis-1,1-dimethylethyl-6-cyanomethyl-2,2-dimethyl-1,3-dioxane-4-acetate, pubchem5924, 3r,5r-6-cyano-3,5-isopropylidenebisoxy hexanoic acid tert-butyl ester, 4r,3r-tert-butyl-6-cyanomethyl-2,2-dimethyl-1,3-dioxane-4-acetate, 4r,6r-tert-butyl-6-cyanomethyl-2,2-dimethyl-1,3-dioxane-4acetate
Solubility Information Solubility in water: .. Other solubilities: soluble in chloroform
InChI Key DPNRMEJBKMQHMC-GHMZBOCLSA-N
SMILES CC(C)(C)OC(=O)C[C@H]1C[C@@H](CC#N)OC(C)(C)O1
IUPAC Name tert-butyl 2-[(4R,6R)-6-(cyanomethyl)-2,2-dimethyl-1,3-dioxan-4-yl]acetate
Molecular Weight (g/mol) 269.34
PubChem CID 2734287
Formula Weight 269.34
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material (4R,6R)-tert-Butyl-6-cyanomethyl-2,2-dimethyl-1,3-dioxane-4-acetate, 97%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0161887 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number
Product Identifier
  • (4R,6R)-tert-Butyl-6-cyanomethyl-2,2-dimethyl-1,3-dioxane-4-acetate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
(4R,6R)-tert-Butyl-6-cyanomethyl-2,2-dimethyl-1,3-dioxane-4-acetate, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.