Learn More
5-(3-Pyridinyloxy)-2-furoic acid, ≥97%, Thermo Scientific™
27.40 € - 185.00 €
Chemical Identifiers
CAS | 852180-39-3 |
---|---|
Molecular Formula | C10H7NO4 |
Molecular Weight (g/mol) | 205.169 |
MDL Number | MFCD03407385 |
InChI Key | ZTYMSWYWSCYFAF-UHFFFAOYSA-N |
Synonym | 5-3-pyridinyloxy-2-furoic acid, 5-pyridin-3-yloxy furan-2-carboxylic acid, 2-furancarboxylic acid,5-3-pyridinyloxy, 5-3-pyridyloxy furan-2-carboxylic acid, 5-3-pyridinyl-oxy-2-furoic acid, 5-pyridin-3-yl oxy furan-2-carboxylic acid |
PubChem CID | 7060545 |
IUPAC Name | 5-pyridin-3-yloxyfuran-2-carboxylic acid |
SMILES | C1=CC(=CN=C1)OC2=CC=C(O2)C(=O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10443253
|
Thermo Scientific Maybridge
AW01246CB |
250 mg |
27.40 €
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10472903
|
Thermo Scientific Maybridge
AW01246DA |
1 g |
91.80 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10148694
|
Thermo Scientific Maybridge
AW01246DE |
5 g |
185.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beschreibung
Chemische Identifikatoren
852180-39-3 | |
205.169 | |
ZTYMSWYWSCYFAF-UHFFFAOYSA-N | |
7060545 | |
C1=CC(=CN=C1)OC2=CC=C(O2)C(=O)O |
C10H7NO4 | |
MFCD03407385 | |
5-3-pyridinyloxy-2-furoic acid, 5-pyridin-3-yloxy furan-2-carboxylic acid, 2-furancarboxylic acid,5-3-pyridinyloxy, 5-3-pyridyloxy furan-2-carboxylic acid, 5-3-pyridinyl-oxy-2-furoic acid, 5-pyridin-3-yl oxy furan-2-carboxylic acid | |
5-pyridin-3-yloxyfuran-2-carboxylic acid |
Spezifikation
852180-39-3 | |
100.0 | |
Amber Glass Bottle | |
MFCD03407385 | |
ZTYMSWYWSCYFAF-UHFFFAOYSA-N | |
5-pyridin-3-yloxyfuran-2-carboxylic acid | |
7060545 | |
97% |
96.5 | |
97% | |
C10H7NO4 | |
5-3-pyridinyloxy-2-furoic acid, 5-pyridin-3-yloxy furan-2-carboxylic acid, 2-furancarboxylic acid,5-3-pyridinyloxy, 5-3-pyridyloxy furan-2-carboxylic acid, 5-3-pyridinyl-oxy-2-furoic acid, 5-pyridin-3-yl oxy furan-2-carboxylic acid | |
C1=CC(=CN=C1)OC2=CC=C(O2)C(=O)O | |
205.169 | |
205.17 | |
5-(3-Pyridinyloxy)-2-furoic acid |
Sicherheit und Handhabung
GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger