missing translation for 'onlineSavingsMsg'
Learn More

5-(4-Fluorophenyl)isoxazole, 99%, Thermo Scientific™

32.50 € - 285.00 €

Chemical Identifiers

CAS 138716-37-7
Molecular Formula C9H6FNO
Molecular Weight (g/mol) 163.151
MDL Number MFCD02183544
InChI Key JZCBQTRPILCIBC-UHFFFAOYSA-N
Synonym 5-4-fluorophenyl isoxazole, 5-4-fluorophenyl-1,2-oxazole, acmc-1c1fl, 1-fluoro-4-isoxazol-5-yl benzene, 1-fluoro-4-1,2-oxazol-5-yl benzene
PubChem CID 2774719
IUPAC Name 5-(4-fluorophenyl)-1,2-oxazole
SMILES C1=CC(=CC=C1C2=CC=NO2)F
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11346589
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H26188.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11356589
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H26188.06
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11366589
View Documents Promotion Details
Thermo Scientific Alfa Aesar
H26188.14
25 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

138716-37-7
163.151
JZCBQTRPILCIBC-UHFFFAOYSA-N
2774719
C1=CC(=CC=C1C2=CC=NO2)F
C9H6FNO
MFCD02183544
5-4-fluorophenyl isoxazole, 5-4-fluorophenyl-1,2-oxazole, acmc-1c1fl, 1-fluoro-4-isoxazol-5-yl benzene, 1-fluoro-4-1,2-oxazol-5-yl benzene
5-(4-fluorophenyl)-1,2-oxazole

Specifications

138716-37-7
C9H6FNO
5-4-fluorophenyl isoxazole, 5-4-fluorophenyl-1,2-oxazole, acmc-1c1fl, 1-fluoro-4-isoxazol-5-yl benzene, 1-fluoro-4-1,2-oxazol-5-yl benzene
C1=CC(=CC=C1C2=CC=NO2)F
163.151
163.15
5-(4-Fluorophenyl)isoxazole
53°C to 55°C
MFCD02183544
JZCBQTRPILCIBC-UHFFFAOYSA-N
5-(4-fluorophenyl)-1,2-oxazole
2774719
99%
Videos
Safety and Handling

Safety and Handling

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications