missing translation for 'onlineSavingsMsg'
Learn More

5,6-Dimethyl-1,10-phenanthroline, 98%, Thermo Scientific Chemicals

90.00 € - 299.00 €

Chemical Identifiers

CAS 3002-81-1
Molecular Formula C14H12N2
Molecular Weight (g/mol) 208.264
MDL Number MFCD00004983
InChI Key BRPQDJPJBCQFSR-UHFFFAOYSA-N
Synonym 1,10-phenanthroline, 5,6-dimethyl, acmc-1afjl, brpqdjpjbcqfsr-uhfffaoysa, 5,6-dimethylpyridino 3,2-h quinoline, 5,6-dimethyl-1,10-phenanthroline
PubChem CID 76357
IUPAC Name 5,6-dimethyl-1,10-phenanthroline
SMILES CC1=C2C=CC=NC2=C3C(=C1C)C=CC=N3
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15403257
View Documents Promotion Details
Thermo Scientific Alfa Aesar
B23717.MD
250 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11319819
View Documents Promotion Details
Thermo Scientific Alfa Aesar
B23717.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

3002-81-1
208.264
BRPQDJPJBCQFSR-UHFFFAOYSA-N
76357
CC1=C2C=CC=NC2=C3C(=C1C)C=CC=N3
C14H12N2
MFCD00004983
1,10-phenanthroline, 5,6-dimethyl, acmc-1afjl, brpqdjpjbcqfsr-uhfffaoysa, 5,6-dimethylpyridino 3,2-h quinoline, 5,6-dimethyl-1,10-phenanthroline
5,6-dimethyl-1,10-phenanthroline

Specifications

3002-81-1
C14H12N2
160918
1,10-phenanthroline, 5,6-dimethyl, acmc-1afjl, brpqdjpjbcqfsr-uhfffaoysa, 5,6-dimethylpyridino 3,2-h quinoline, 5,6-dimethyl-1,10-phenanthroline
CC1=C2C=CC=NC2=C3C(=C1C)C=CC=N3
208.264
208.26
5,6-Dimethyl-1,10-phenanthroline
266°C to 269°C
MFCD00004983
Hygroscopic
BRPQDJPJBCQFSR-UHFFFAOYSA-N
5,6-dimethyl-1,10-phenanthroline
76357
98%
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 221-100-2

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only