missing translation for 'onlineSavingsMsg'
Learn More
5-Amino-2,4,6-triiodoisophthalic acid, 95%, Thermo Scientific Chemicals Product Code.: 11429717

5-Amino-2,4,6-triiodoisophthalic acid, 95%, Thermo Scientific Chemicals

Product Code. 11429717
50 g
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy

Product Code. 11429717

Brand: Thermo Scientific Alfa Aesar B22178.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35453-19-1
Molecular Formula C8H4I3NO4
Molecular Weight (g/mol) 558.836
MDL Number MFCD00190167
InChI Key JEZJSNULLBSYHV-UHFFFAOYSA-N
Synonym 5-amino-2,4,6-triiodoisophthalic acid, unii-a17g990rt0, 5-amino-2,4,6-triiodo-1,3-benzenedicarboxylic acid, 5-amino-2,4,6-iodoisophthalic acid, 1,3-benzenedicarboxylic acid, 5-amino-2,4,6-triiodo, 5-amino-2,4,6-triiodoisophthalicacid, isophthalic acid, 5-amino-2,4,6-triiodo, acmc-209ih0, ksc224s6b, aronis25361Show MoreShow Less
PubChem CID 3015783
IUPAC Name 5-amino-2,4,6-triiodobenzene-1,3-dicarboxylic acid
SMILES C1(=C(C(=C(C(=C1I)N)I)C(=O)O)I)C(=O)O

Specifications

Melting Point 265°C to 270°C
Quantity 50 g
Sensitivity Light and moisture sensitive
Formula Weight 558.84
Percent Purity 95%
Chemical Name or Material 5-Amino-2,4,6-triiodoisophthalic acid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10230760 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberB22178.18,B22178.09,B22178.30
Lot Number10230760 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB22178.18,B22178.09,B22178.30
Lot Number10220006 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB22178.18,B22178.09,B22178.30
Lot Number5001J10X Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB22178.18,B22178.09,B22178.30
Lot Number5001N21S Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB22178.18,B22178.09,B22178.30

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
5-Amino-2,4,6-triiodoisophthalic acid, 95%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.