missing translation for 'onlineSavingsMsg'
Learn More
5-Aminoisophthalic acid, 98%, Thermo Scientific Chemicals Product Code.: 10459500

5-Aminoisophthalic acid, 98%, Thermo Scientific Chemicals

Product Code. 10459500
100 g, Plastic bottle
Click to view available options
Quantity:
25 g
100 g
Packaging:
Plastic bottle
This item is not returnable. View return policy

Product Code. 10459500

Brand: Thermo Scientific Acros 181331000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99-31-0
Molecular Formula C8H7NO4
Molecular Weight (g/mol) 181.15
MDL Number MFCD00002522
InChI Key KBZFDRWPMZESDI-UHFFFAOYSA-N
Synonym 5-aminoisophthalic acid, 1,3-benzenedicarboxylic acid, 5-amino, 5-amino-1,3-isophthalic acid, isophthalic acid, 5-amino, 5-amino-1,3-benzenedicarboxylic acid, 3,5-dicarboxyaniline, 5-aminoisophtalic acid, 3,5-dicarboxy-aniline, pubchem20539, anhydrousShow MoreShow Less
PubChem CID 66833
IUPAC Name 5-aminobenzene-1,3-dicarboxylic acid
SMILES C1=C(C=C(C=C1C(=O)O)N)C(=O)O

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point >300°C
Color White to Cream
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Plastic bottle
Quantity 100 g
Beilstein 14, I, 636
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethanol
Formula Weight 181.15
Percent Purity 98%
Physical Form Granular Powder
Chemical Name or Material 5-Aminoisophthalic acid, 98+%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0466121 Certificate TypeCertificate of Analysis Date24/09/2024 Catalog Number181331000,181330250
Lot NumberA0452259 Certificate TypeCertificate of Analysis Date17/04/2024 Catalog Number181331000,181330250
Lot NumberA0414505 Certificate TypeCertificate of Analysis Date12/12/2022 Catalog Number181331000,181330250
Lot NumberA0424050 Certificate TypeCertificate of Analysis Date16/11/2022 Catalog Number181331000,181330250
Lot NumberA0440743 Certificate TypeCertificate of Analysis Date08/04/2022 Catalog Number
compliance-icons
Product Identifier
  • 5-Aminoisophthalic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
5-Aminoisophthalic acid, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.