missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Bromo-1-methylimidazole, 97%, Thermo Scientific Chemicals
138.69 € - 471.35 €
Chemical Identifiers
CAS | 1003-21-0 |
---|---|
Molecular Formula | C4H5BrN2 |
Molecular Weight (g/mol) | 161.00 |
MDL Number | MFCD01632218 |
InChI Key | HATLLUIOEIXWGD-UHFFFAOYSA-N |
Synonym | 5-bromo-1-methyl-1h-imidazole, 5-bromo-n-methylimidazole, pubchem8951, 1h-imidazole, 5-bromo-1-methyl, 5-bromo-1-methyl-imidazole, imidazole,5-bromo-1-methyl, 5-bromo-n-methyl-imidazole, 5-bromo-1-methyl imidazole, acmc-1c6k3, 1-methyl-5-bromo-1h-imidazole |
PubChem CID | 1515258 |
IUPAC Name | 5-bromo-1-methylimidazole |
SMILES | CN1C=NC=C1Br |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10784955
|
Thermo Scientific Acros
381040050 |
5 g |
138.69 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10794955
|
Thermo Scientific Acros
381040250 |
25 g |
471.35 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1003-21-0 | |
161.00 | |
HATLLUIOEIXWGD-UHFFFAOYSA-N | |
1515258 | |
CN1C=NC=C1Br |
C4H5BrN2 | |
MFCD01632218 | |
5-bromo-1-methyl-1h-imidazole, 5-bromo-n-methylimidazole, pubchem8951, 1h-imidazole, 5-bromo-1-methyl, 5-bromo-1-methyl-imidazole, imidazole,5-bromo-1-methyl, 5-bromo-n-methyl-imidazole, 5-bromo-1-methyl imidazole, acmc-1c6k3, 1-methyl-5-bromo-1h-imidazole | |
5-bromo-1-methylimidazole |
Specifications
1003-21-0 | |
120°C (15.0 mmHg) | |
96% min. (GC) | |
C4H5BrN2 | |
5-bromo-1-methyl-1h-imidazole, 5-bromo-n-methylimidazole, pubchem8951, 1h-imidazole, 5-bromo-1-methyl, 5-bromo-1-methyl-imidazole, imidazole,5-bromo-1-methyl, 5-bromo-n-methyl-imidazole, 5-bromo-1-methyl imidazole, acmc-1c6k3, 1-methyl-5-bromo-1h-imidazole | |
CN1C=NC=C1Br | |
161.00 | |
161 | |
5-Bromo-1-methylimidazole |
40°C to 41°C | |
82°C | |
Glass bottle | |
MFCD01632218 | |
HATLLUIOEIXWGD-UHFFFAOYSA-N | |
5-bromo-1-methylimidazole | |
1515258 | |
97% |
Safety and Handling
GHS Signal Word: Danger
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- 5-Bromo-1-methylimidazole
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only