Learn More
5-Bromo-5-nitro-1,3-dioxane, 98%, Thermo Scientific Chemicals
Description
5-Bromo-5-nitro-1,3-dioxane is used as a stabilizer and preserving agent for biological molecules and solutions such as antibodies and antisera. Bronidox is used in a variety of rinse-off cosmetic. It can be used alone or in combination with methylisothiazolone.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
• 5-Bromo-5-nitro-1,3-dioxane is a chemical compound showing antimicrobial activity against gram-negative and gram-positive bacteria, yeast, and fungi
• This compound is capable of promoting the oxidation of essential protein thiols, causing inhibition of enzyme activity leading to the inhibition of microbial growth
Application
• 5-Bromo-5-nitro-1,3-dioxane has been used as a stabilizer and preserving agent for biological molecules and solutions, including antibodies and antisera
• It can be used alone or combined with methylisothiazolinone, which is also considered to be an effective preservative

Specifications
Specifications
Melting Point | 58°C to 61°C |
Color | White |
Quantity | 25 g |
Solubility Information | Soluble in water at 12.5mg/ml |
Formula Weight | 212 |
Percent Purity | 98% |
Physical Form | Powder |
Chemical Name or Material | 5-Bromo-5-nitro-1,3-dioxane |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
P10L002 | Certificate of Analysis | 12/03/2025 | J65443.14,J65443.06 |
M28L052 | Certificate of Analysis | 30/01/2025 | J65443.14,J65443.06 |
Z24K034 | Certificate of Analysis | 23/01/2025 | J65443.14,J65443.06 |
10252980 | Certificate of Analysis | 13/11/2024 | J65443.14,J65443.06 |
X09K052 | Certificate of Analysis | 18/10/2024 | J65443.14,J65443.06 |
Safety and Handling



- 5-Bromo-5-nitro-1,3-dioxane
- Danger
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 A
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : 5-Bromo-5-nitro-1,3-dioxane
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.