Learn More
Tocris Bioscience™ (±)-5'-Chloro-5'-deoxy-ENBA
Highly selective A1 agonist
Brand:  Tocris Bioscience™ 3576/50
970.33 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
 View more versions of this product
Product Code. 18004542
  • 1200.00 € / 50mg
Explore more special offers
Description
Description
(±)-5'-Chloro-5'-deoxy-ENBA is a highly selective adenosine A1 receptor agonist (Ki values are 0.51, 1290, 1340 and 2740 nM at A1, A3, A2A and A2B receptors respectively). Reverses formalin-induced nocifensive behavior in mice; antinociceptive.
Specifications
Specifications
>98%
(±)-5'-Chloro-5'-deoxy-ENBA
(±)-5'-Chloro-5'-deoxy-ENBA
50 mg
379.84