missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ (±)-5'-Chloro-5'-deoxy-ENBA
Highly selective A1 agonist
283.00 € - 1200.00 €
Specifications
Purity | >98% |
---|---|
Product Type | (±)-5'-Chloro-5'-deoxy-ENBA |
Molecular Weight (g/mol) | 379.84 |
Inhibitors | (±)-5'-Chloro-5'-deoxy-ENBA |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18044902
|
Tocris Bioscience™
3576/10 |
10 mg |
283.00 €
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18004542
|
Tocris Bioscience™
3576/50 |
50 mg |
1200.00 €
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Highly selective adenosine A1 receptor agonist (Ki values are 0.51, 1290, 1340 and 2740 nM at A1, A3, A2A and A2B receptors respectively). Reverses formaline-induced nocifensive behavior in mice; antinociceptive.Specifications
>98% | |
(±)-5'-Chloro-5'-deoxy-ENBA | |
379.84 | |
(±)-5'-Chloro-5'-deoxy-ENBA |
For Research Use Only